Search
Search

MAGI2
MAGI2-AS2GCTAAAAAATACAGAGTTCTAAAAA[G/A]TGTACAATAAATGTTATCTATTGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
|||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGI2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
| MAGI2 - membrane associated guanylate kinase, WW and PDZ domain containing 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001301128.1 | Intron | NP_001288057.1 | ||||
| NM_012301.3 | Intron | NP_036433.2 | ||||
| XM_011516718.1 | Intron | XP_011515020.1 | ||||
| XM_011516719.2 | Intron | XP_011515021.1 | ||||
| XM_011516720.2 | Intron | XP_011515022.1 | ||||
| XM_011516726.2 | Intron | XP_011515028.1 | ||||
| XM_011516728.1 | Intron | XP_011515030.1 | ||||
| XM_017012840.1 | Intron | XP_016868329.1 | ||||
| XM_017012841.1 | Intron | XP_016868330.1 | ||||
| XM_017012842.1 | Intron | XP_016868331.1 | ||||
| XM_017012843.1 | Intron | XP_016868332.1 | ||||
| XM_017012844.1 | Intron | XP_016868333.1 | ||||
| XM_017012845.1 | Intron | XP_016868334.1 | ||||
| XM_017012846.1 | Intron | XP_016868335.1 | ||||
| XM_017012847.1 | Intron | XP_016868336.1 | ||||
| XM_017012848.1 | Intron | XP_016868337.1 | ||||
| XM_017012849.1 | Intron | XP_016868338.1 | ||||
| XM_017012850.1 | Intron | XP_016868339.1 | ||||
| XM_017012851.1 | Intron | XP_016868340.1 | ||||
| XM_017012852.1 | Intron | XP_016868341.1 | ||||
| MAGI2-AS2 - MAGI2 antisense RNA 2 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||