Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTATAGATGTCTTTCTCTCTGTGT[A/G]TATATATATATATATATACACACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 143450 | ||||||||||||||||||||
Literature Links: |
HADHB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HADHB - hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000183.2 | Intron | NP_000174.1 | ||||
NM_001281512.1 | Intron | NP_001268441.1 | ||||
NM_001281513.1 | Intron | NP_001268442.1 | ||||
XM_011532803.1 | Intron | XP_011531105.1 | ||||
XM_017003939.1 | Intron | XP_016859428.1 |