Search
Search

OTUD7BGAAGCCAAGCATTAGAAGATGTTTA[T/C]TACTATCATCATGGGCCATACCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTUD7B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| OTUD7B - OTU deubiquitinase 7B | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_020205.3 | Intron | NP_064590.2 | ||||
| XM_011509782.1 | Intron | XP_011508084.1 | ||||
| XM_011509783.2 | Intron | XP_011508085.1 | ||||
| XM_011509784.2 | Intron | XP_011508086.1 | ||||
| XM_011509785.2 | Intron | XP_011508087.1 | ||||
| XM_011509786.1 | Intron | XP_011508088.1 | ||||
| XM_011509787.1 | Intron | XP_011508089.1 | ||||
| XM_011509788.1 | Intron | XP_011508090.1 | ||||
| XM_017001850.1 | Intron | XP_016857339.1 | ||||
| XM_017001851.1 | Intron | XP_016857340.1 | ||||