Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACACAATATGTAAATAAATGGGT[C/T]CGTTACATTTCAATAAAACTTTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608401 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6503 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6503 - microRNA 6503 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MS4A4E - membrane spanning 4-domains A4E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011545416.2 | Intron | XP_011543718.1 | ||||
XM_011545417.2 | Intron | XP_011543719.1 | ||||
XM_011545418.2 | Intron | XP_011543720.1 | ||||
XM_011545419.2 | Intron | XP_011543721.1 | ||||
XM_017018641.1 | Intron | XP_016874130.1 |
Set Membership: |
HapMap |