Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1F10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | ||||||
|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
| IL1F10 - interleukin 1 family member 10 (theta) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| IL36RN - interleukin 36 receptor antagonist | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_012275.2 | 1547 | UTR 3 | NP_036407.1 | |||
| NM_173170.1 | 1547 | UTR 3 | NP_775262.1 | |||
Set Membership: |
|