Search
Search

Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGUOK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| DGUOK - deoxyguanosine kinase | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001318859.1 | Intron | NP_001305788.1 | ||||
| NM_001318860.1 | Intron | NP_001305789.1 | ||||
| NM_001318861.1 | Intron | NP_001305790.1 | ||||
| NM_001318862.1 | Intron | NP_001305791.1 | ||||
| NM_001318863.1 | Intron | NP_001305792.1 | ||||
| NM_080916.2 | Intron | NP_550438.1 | ||||
| NM_080918.2 | Intron | NP_550440.1 | ||||
| XM_011532647.2 | Intron | XP_011530949.1 | ||||