Search
Search

LOC105370854
TLN2TTGTCCGTGCAGCCCAGAAGGCAGC[A/T]TTTGGCAAAGCTGATGACGACGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105370854 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||||||||
|---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
| LOC105370854 - uncharacterized LOC105370854 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| TLN2 - talin 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_015059.2 | 7753 | Silent Mutation | GCA,GCT | A,A 2479 | NP_055874.2 | |
| XM_005254708.4 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254765.1 | |
| XM_005254710.4 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254767.1 | |
| XM_005254711.4 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254768.1 | |
| XM_005254712.4 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254769.1 | |
| XM_005254713.4 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254770.1 | |
| XM_005254714.3 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254771.1 | |
| XM_005254715.2 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_005254772.1 | |
| XM_006720717.3 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_006720780.1 | |
| XM_017022665.1 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_016878154.1 | |
| XM_017022666.1 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_016878155.1 | |
| XM_017022667.1 | 7753 | Silent Mutation | GCA,GCT | A,A 2494 | XP_016878156.1 | |
| XM_017022668.1 | 7753 | Silent Mutation | GCA,GCT | A,A 2479 | XP_016878157.1 | |
| XM_017022669.1 | 7753 | Silent Mutation | GCA,GCT | A,A 2449 | XP_016878158.1 | |