Search
Search

| Assay Name: | rno-miR-743a |
|---|---|
| miRBase Accession Number: | MI0006162 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAAAGACGCCAAACUGGGUAGA |
| Species: | Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | GAAAGACGCCAAACUGGGUAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-743a-3p | MIMAT0005334 |