TaqMan® OpenArray® Rodent MicroRNA Panel
Das tatsächliche Produkt kann abweichen
Applied Biosystems™
TaqMan® OpenArray® Rodent MicroRNA Panel
Have Questions?
Katalognummer 4461105
Preis (EUR)
1.316,00
1 panel
Preis (EUR)
1.316,00
1 panel
TaqMan® OpenArray® MicroRNA Panels provide many of the advantages of TaqMan® MicroRNA Assays in a high-throughput plate and are ideal for medium to large miRNA profiling studies. TaqMan® OpenArray® MicroRNA Panels enable the running of hundreds of TaqMan® MicroRNA Assays in parallel per sample. This greatly simplifies the generation of a comprehensive miRNA profile for up to 36 samples in a single working day.
Key product features:
• Comprehensive coverage-generate miRNA profiles across 750 rodent (mouse and rat) well-characterized miRNAs with as little as 100 nanograms of total RNA
• High throughput-run up to 36 samples per 8-hour working day with minimal hands-on time and no need for robotics
• Cost effective-3 samples can be run per panel in 33 nanoliter reaction volumes resulting in significant reagent cost savings
Provides Comprehensive Coverage of Well-Studied miRNAs in Sanger miRBase
Many well-characterized rodent (mouse and rat) miRNA sequences from the Sanger miRBase are included. The content is derived from miRBase v15, providing close to full coverage of Sanger well-studied miRNAs using highly-validated TaqMan® MicroRNA Assays.
Rodent
Megaplex™ Primer PoolsMicroRNA TargetsControl (+ve)Control (-ve)
A37551
B37551
Total750
Streamlined Workflow and High Throughput Sample Handling
The workflow is completed in five steps (Fig 1):
1. Split total RNA sample and process in parallel using Megaplex™ Primer Pools A and B and the supporting TaqMan® MicroRNA Reverse Transcription Kit.
2. Resulting cDNA is preamplified prior to real-time PCR analysis using Megaplex™ PreAmp Pools and TaqMan® PreAmp Master Mix.
3. Preamplified cDNA is diluted, mixed 1:1 with TaqMan® OpenArray® Real-Time PCR Master Mix in the 384-well OpenArray® Sample Loading Plate.
4. TaqMan® OpenArray® Rodent MicroRNA Panels are automatically loaded using the AccuFill™ System.
5. Up to 3 panels are cycled simultaneously on an OpenArray® Real-Time PCR Instrument.
High Concordance to Single-tube Assays
The TaqMan® OpenArray® Rodent MicroRNA Panel has demonstrated high concordance with an R2 value of 0.97 to the mostly commonly used tool for validation: individual TaqMan MicroRNA Assays (Fig 2). The same panel is used in the TaqMan® OpenArray® Rodent MicroRNA Panel, QuantStudio™ 12K Flex, enabling accurate answers to help minimize validation.
Additional Required Reagents
These products are also required to run the TaqMan® OpenArray® Rodent MicroRNA Panel:
• Megaplex™ Primer Pools (PN 4444750 or 4444766)
• TaqMan® MicroRNA Reverse Transcription Kit (PN 4366596 or 4366597)
• TaqMan® PreAmp Master Mix (PN 4391128)
• TaqMan® OpenArray® Real-Time PCR Master Mix (PN 4462159 or 4462164)
• OpenArray® Real-Time Accessories Kit (PN 4453975)
• OpenArray® 384-well Sample Plates (PN 4406947)
• OpenArray® AccuFill™ System Tips (PN 4457246 or 4458107)
Download OpenArray® Plate Files
OpenArray® Plate Files (TPF) are needed to process your TaqMan® OpenArray® MicroRNA Panels on the OpenArray® Real-Time PCR System.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 panel
Inhalt und Lagerung
Store at or below –20°C. Do not store in a frost-free freezer.