Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002768
          Assay Name: hsa-miR-1233
          miRBase Accession Number: MI0006323
          miRBase Version: v22.1
          Chromosome Location: Chr.15: 34382069 - 34382150 [-] on Build GRCh38
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Species: Human, Pan troglodytes, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002768
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000578, mir-1233

          Mature miRNA Sequence:

          UGAGCCCUGUCCUCCCGCAG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-1233-3p MIMAT0005588
          Pan troglodytes ptr-miR-1233 MIMAT0007966
          Rhesus monkey mml-miR-1233 MIMAT0012789

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-1233-1
          Stem-loop Accession # MI0006323
          Stem-loop Sequence
          GUGAGUGGGAGGCCAGGGCACGGCAGGGGGAGCUGCAGGGCUAUGGGAGGGGCCCCAGCGUCUGAGCCCUGUCCUCCCGCAG
          Chromosome Location Chr. 15 - 34382069 - 34382150 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005588
          miRBase ID: hsa-miR-1233-3p
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Chromosome Location: Chr. 15 - 34528290 - 34528371 [-] on Build GRCh38

          Stem-loop ID hsa-mir-1233-2
          Stem-loop Accession # MI0015973
          Stem-loop Sequence
          GUGAGUGGGAGGCCAGGGCACGGCAGGGGGAGCUGCAGGGCUAUGGGAGGGGCCCCAGCGUCUGAGCCCUGUCCUCCCGCAG
          Chromosome Location Chr. 15 - 34528290 - 34528371 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0005588
          miRBase ID: hsa-miR-1233-3p
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Chromosome Location: Chr. 15 - 34528290 - 34528371 [-] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-1233-1
          Stem-loop Accession # MI0008434
          Stem-loop Sequence
          GUGAGUGGGAGGCCAGGGCAUGGCAGGGGGAGCUGCAGGGCCGUCGGAGGGGCCCCAGCAUCUGAGCCCUGUCCUCCCGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007966
          miRBase ID: ptr-miR-1233
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1233-2
          Stem-loop Accession # MI0008435
          Stem-loop Sequence
          GUGAGUGGGAGGCCAGUGCACGGCAGGGGGAGCUGCAGGGCCGUCGGAGGGGCCCCAGCGUCUGAGCCCUGUCCUCCCGCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007966
          miRBase ID: ptr-miR-1233
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Chromosome Location: NA

          Stem-loop ID ptr-mir-1233-3
          Stem-loop Accession # MI0008436
          Stem-loop Sequence
          UGAGUGGGAGGCCAGUGCACGGCAGGGGGAGCUGCAGGGCCGUCGGAGGGGCCCCAGCGUCUGAGCCCUGUCCUCCCGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007966
          miRBase ID: ptr-miR-1233
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-1233
          Stem-loop Accession # MI0012546
          Stem-loop Sequence
          GUGAGUGGGAGGCCAGGGCAGGGCAGGGGGAGCUGCAGGGCCUUCGGAGGGGCCCCAGCGUCUGAGCCCUGUCCUCCCGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012789
          miRBase ID: mml-miR-1233
          Mature miRNA Sequence: UGAGCCCUGUCCUCCCGCAG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Human Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03305411_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13610
          Ambion® Pre-miR™ miRNA Precursor : PM13610
          mirVana® miRNA inhibitor : MH13610
          mirVana® miRNA mimic : MC13610
          TaqMan™ Advanced miRNA Assay : 477877_mir
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          FGMIR : TaqManTM Human MicroRNA Array B v3
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-miR-1233 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline