Search
Search

| Assay Name: | hsa-miR-1233-3p |
|---|---|
| miRBase Accession Number: | MI0006323 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGAGCCCUGUCCUCCCGCAG |
| Species: | Human, Pan troglodytes, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000578, mir-1233 |
Mature miRNA Sequence: | UGAGCCCUGUCCUCCCGCAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1233-3p | MIMAT0005588 |
| Pan troglodytes | ptr-miR-1233 | MIMAT0007966 |
| Rhesus monkey | mml-miR-1233 | MIMAT0012789 |