Search
Search

| Assay Name: | mmu-miR-107* |
|---|---|
| miRBase Accession Number: | MI0000684 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCUUCUUUACAGUGUUGCCUUG |
| Species: | Mouse, Chicken, Columba livia, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Xenopus laevis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | AGCUUCUUUACAGUGUUGCCUUG |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-107-5p | MIMAT0017048 | |
| Chicken | gga-miR-107-5p | MIMAT0026520 | |
| Columba livia | cli-miR-107-5p | MIMAT0038489 | |
| Dasypus novemcinctus | dno-miR-107-5p | MIMAT0047608 | |
| Guinea pig | cpo-miR-107-5p | MIMAT0046959 | |
| Pteropus alecto | pal-miR-107a-5p | MIMAT0040201 | |
| View More Rows | |||
| Python bivittatus | pbv-miR-107-5p | MIMAT0038907 | |
| Rabbit | ocu-miR-107-5p | MIMAT0048199 | |
| Rhesus monkey | mml-miR-107-5p | MIMAT0026584 | |
| Salmo salar | ssa-miR-107-5p | MIMAT0032290 | |
| Xenopus laevis | xla-miR-107-5p | MIMAT0046420 | |