Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH19864
          Assay Name: mmu-miR-107-5p
          miRBase Accession Number: MI0000684
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 34820687 - 34820773 [-] on Build GRCm38
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Species: Mouse, Chicken, Columba livia, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Xenopus laevis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH19864

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000024, mir-103

          Mature miRNA Sequence:

          AGCUUCUUUACAGUGUUGCCUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-107-5p MIMAT0017048
          Chicken gga-miR-107-5p MIMAT0026520
          Columba livia cli-miR-107-5p MIMAT0038489
          Dasypus novemcinctus dno-miR-107-5p MIMAT0047608
          Guinea pig cpo-miR-107-5p MIMAT0046959
          Pteropus alecto pal-miR-107a-5p MIMAT0040201
          View More Rows
          Python bivittatus pbv-miR-107-5p MIMAT0038907
          Rabbit ocu-miR-107-5p MIMAT0048199
          Rhesus monkey mml-miR-107-5p MIMAT0026584
          Salmo salar ssa-miR-107-5p MIMAT0032290
          Xenopus laevis xla-miR-107-5p MIMAT0046420

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-107
          Stem-loop Accession # MI0000684
          Stem-loop Sequence
          UUCUCUGUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC
          Chromosome Location Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017048
          miRBase ID: mmu-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38

          Chicken

          Stem-loop ID gga-mir-107
          Stem-loop Accession # MI0001215
          Stem-loop Sequence
          CUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026520
          miRBase ID: gga-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-107
          Stem-loop Accession # MI0029952
          Stem-loop Sequence
          GUGUUCUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038489
          miRBase ID: cli-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-107
          Stem-loop Accession # MI0038962
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGAAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUAGAGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047608
          miRBase ID: dno-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-107
          Stem-loop Accession # MI0038620
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGAUCAAGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046959
          miRBase ID: cpo-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-107a
          Stem-loop Accession # MI0032547
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040201
          miRBase ID: pal-miR-107a-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-107
          Stem-loop Accession # MI0030209
          Stem-loop Sequence
          CUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUAGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038907
          miRBase ID: pbv-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-107
          Stem-loop Accession # MI0039274
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048199
          miRBase ID: ocu-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-107
          Stem-loop Accession # MI0002754
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026584
          miRBase ID: mml-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-107
          Stem-loop Accession # MI0026441
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGAUCAAGCAGCAUUGUACAGGGCUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032290
          miRBase ID: ssa-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-107
          Stem-loop Accession # MI0038232
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046420
          miRBase ID: xla-miR-107-5p
          Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307054_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM19864
          Ambion® Pre-miR™ miRNA Precursor : PM19864
          TaqMan™ MicroRNA Assay : 465082_mat
          mirVana® miRNA mimic : MC19864


          miRBase Alias:

          Mouse: mmu-miR-107* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline