Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm03307054_pri
Assay Name:
mmu-mir-107
Stem-loop ID:
mmu-mir-107
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.19: 34820687 - 34820773 [-] on Build GRCm38
Stem-loop Location:
Chr.19: 34820687 - 34820773 [-] on Build GRCm38
Mature miRNA ID:
mmu-miR-107-5p
mmu-miR-107-3p
Species:
Mouse
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Mm03307054_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
77
Chromosome Location:
Chr. 19 - 34820988 - 34820988 [+] on 34820988 Build GRCm38
Species:
Mouse
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Mouse
mmu-miR-107-5p
MIMAT0017048
Mouse
mmu-miR-107-3p
MIMAT0000647
Stem-loop Details
Mouse
Stem-loop ID
mmu-mir-107
Stem-loop Accession #
MI0000684
Stem-loop Sequence
UUCUCUGUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC
Chromosome Location
Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38
Mature MicroRNA
miRBase Accession #: MIMAT0017048
miRBase ID: mmu-miR-107-5p
Mature miRNA Sequence: AGCUUCUUUACAGUGUUGCCUUG
Chromosome Location: Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38
miRBase Accession #: MIMAT0000647
miRBase ID: mmu-miR-107-3p
Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
Chromosome Location: Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38
Gene Family ID
MIPF0000024,
mir-103
Distance to Amplicon
181 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM19864
Ambion
®
Pre-miR
™
miRNA Precursor :
PM19864
TaqMan
™
MicroRNA Assay :
465082_mat
mir
Vana
®
miRNA inhibitor :
MH19864
mir
Vana
®
miRNA mimic :
MC19864
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM10056
Ambion
®
Pre-miR
™
miRNA Precursor :
PM10056
TaqMan
™
MicroRNA Assay :
000443
mir
Vana
®
miRNA inhibitor :
MH10056
mir
Vana
®
miRNA mimic :
MC10056
TaqMan
™
Advanced miRNA Assay :
478254_mir
TaqMan
™
Advanced miRNA Assay :
mmu478254_mir
TaqMan
™
Advanced miRNA Assay :
rno478254_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline