Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10056
          Assay Name: hsa-miR-107
          miRBase Accession Number: MI0000114
          miRBase Version: v22.1
          Chromosome Location: Chr.10: 89592747 - 89592827 [-] on Build GRCh38
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Species: Human, Mouse, Rat, Chicken, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Monodelphis domestica, Ophiophagus hannah, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC10056

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000024, mir-103

          Mature miRNA Sequence:

          AGCAGCAUUGUACAGGGCUAUCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-107 MIMAT0000104
          Mouse mmu-miR-107-3p MIMAT0000647
          Rat rno-miR-107-3p MIMAT0000826
          Chicken gga-miR-107-3p MIMAT0001147
          Dasypus novemcinctus dno-miR-107-3p MIMAT0047609
          Gorilla gorilla ggo-miR-107 MIMAT0002460
          View More Rows
          Guinea pig cpo-miR-107-3p MIMAT0046960
          Horse eca-miR-107b MIMAT0012886
          Lagothrix lagotricha lla-miR-107 MIMAT0002462
          Macaca nemestrina mne-miR-107 MIMAT0002463
          Monodelphis domestica mdo-miR-107 MIMAT0004100
          Ophiophagus hannah oha-miR-107-3p MIMAT0036655
          Pan paniscus ppa-miR-107 MIMAT0002464
          Pan troglodytes ptr-miR-107 MIMAT0002459
          Pig ssc-miR-107 MIMAT0002155
          Pongo pygmaeus ppy-miR-107 MIMAT0002461
          Rabbit ocu-miR-107-3p MIMAT0048200
          Rhesus monkey mml-miR-107-3p MIMAT0002458
          Taeniopygia guttata tgu-miR-107 MIMAT0014556
          Tetraodon nigroviridis tni-miR-107 MIMAT0003044
          Xenopus tropicalis xtr-miR-107 MIMAT0003584
          Zebrafish dre-miR-107a-3p MIMAT0001817

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-107
          Stem-loop Accession # MI0000114
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location Chr. 10 - 89592747 - 89592827 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000104
          miRBase ID: hsa-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: Chr. 10 - 89592747 - 89592827 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-107
          Stem-loop Accession # MI0000684
          Stem-loop Sequence
          UUCUCUGUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC
          Chromosome Location Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000647
          miRBase ID: mmu-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: Chr. 19 - 34820687 - 34820773 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-107
          Stem-loop Accession # MI0000890
          Stem-loop Sequence
          UUCUCUCUGCUUUAAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC
          Chromosome Location Chr. 1 - 253128250 - 253128336 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000826
          miRBase ID: rno-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: Chr. 1 - 253128250 - 253128336 [-] on Build Rnor_6.0

          Chicken

          Stem-loop ID gga-mir-107
          Stem-loop Accession # MI0001215
          Stem-loop Sequence
          CUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001147
          miRBase ID: gga-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-107
          Stem-loop Accession # MI0038962
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGAAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUAGAGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047609
          miRBase ID: dno-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-107
          Stem-loop Accession # MI0002756
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002460
          miRBase ID: ggo-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-107
          Stem-loop Accession # MI0038620
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGAUCAAGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046960
          miRBase ID: cpo-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-107b
          Stem-loop Accession # MI0012643
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012886
          miRBase ID: eca-miR-107b
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-107
          Stem-loop Accession # MI0002758
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002462
          miRBase ID: lla-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-107
          Stem-loop Accession # MI0002759
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002463
          miRBase ID: mne-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-107
          Stem-loop Accession # MI0005286
          Stem-loop Sequence
          CUUCUUUCUGCUUUCGGCUUCUCUACAGUGUUGCCUUGUGGCGUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004100
          miRBase ID: mdo-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-107
          Stem-loop Accession # MI0031316
          Stem-loop Sequence
          CUCUUUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036655
          miRBase ID: oha-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-107
          Stem-loop Accession # MI0002760
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002464
          miRBase ID: ppa-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-107
          Stem-loop Accession # MI0002755
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002459
          miRBase ID: ptr-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-107
          Stem-loop Accession # MI0002449
          Stem-loop Sequence
          UUCUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGAGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002155
          miRBase ID: ssc-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-107
          Stem-loop Accession # MI0002757
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002461
          miRBase ID: ppy-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-107
          Stem-loop Accession # MI0039274
          Stem-loop Sequence
          AGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048200
          miRBase ID: ocu-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-107
          Stem-loop Accession # MI0002754
          Stem-loop Sequence
          CUCUCUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002458
          miRBase ID: mml-miR-107-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-107
          Stem-loop Accession # MI0013773
          Stem-loop Sequence
          UUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCAUGGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014556
          miRBase ID: tgu-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-107
          Stem-loop Accession # MI0003380
          Stem-loop Sequence
          UCUGUCUGCUGUGAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGCGAUCAAGCAGCAUUGUACAGGGCUAUCACCGCACACAGAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003044
          miRBase ID: tni-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-107
          Stem-loop Accession # MI0004823
          Stem-loop Sequence
          CUGCUUUCAGCUUCUUUACAGUGUUGCCUUGUGGCAUGGAGUUCAAGCAGCAUUGUACAGGGCUAUCAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003584
          miRBase ID: xtr-miR-107
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-107a
          Stem-loop Accession # MI0001963
          Stem-loop Sequence
          UCUGUGUGCUCUGAGCUUCUUUACAGUGUUGUCUUGUGGCAUGGAGAUCAAGCAGCAUUGUACAGGGCUAUCACAGCACACUGAACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001817
          miRBase ID: dre-miR-107a-3p
          Mature miRNA Sequence: AGCAGCAUUGUACAGGGCUAUCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465997_pri
          TaqMan™ Pri-miRNA Assay : Hs03302781_pri
          TaqMan™ Pri-miRNA Assay : Mm03307054_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10056
          Ambion® Pre-miR™ miRNA Precursor : PM10056
          TaqMan™ MicroRNA Assay : 000443
          mirVana® miRNA inhibitor : MH10056
          TaqMan™ Advanced miRNA Assay : 478254_mir
          TaqMan™ Advanced miRNA Assay : mmu478254_mir
          TaqMan™ Advanced miRNA Assay : rno478254_mir


          miRBase Alias:

          Mouse: mmu-miR-107 (v17)
          Rat: rno-miR-107 (v18)
          Danio rerio: dre-miR-107 (v18)
          Danio rerio: dre-miR-107a (v20)
          Fugu rubripes: fru-miR-107 (v20)
          Gallus gallus: gga-miR-107 (v19)
          Macaca mulatta: mml-miR-107 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline