Search
Search

| Assay Name: | hsa-miR-107 |
|---|---|
| miRBase Accession Number: | MI0000114 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCAGCAUUGUACAGGGCUAUCA |
| Species: | Human, Mouse, Rat, Chicken, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Lagothrix lagotricha, Macaca nemestrina, Monodelphis domestica, Ophiophagus hannah, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000024, mir-103 |
Mature miRNA Sequence: | AGCAGCAUUGUACAGGGCUAUCA |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-107 | MIMAT0000104 | |
| Mouse | mmu-miR-107-3p | MIMAT0000647 | |
| Rat | rno-miR-107-3p | MIMAT0000826 | |
| Chicken | gga-miR-107-3p | MIMAT0001147 | |
| Dasypus novemcinctus | dno-miR-107-3p | MIMAT0047609 | |
| Gorilla gorilla | ggo-miR-107 | MIMAT0002460 | |
| View More Rows | |||
| Guinea pig | cpo-miR-107-3p | MIMAT0046960 | |
| Horse | eca-miR-107b | MIMAT0012886 | |
| Lagothrix lagotricha | lla-miR-107 | MIMAT0002462 | |
| Macaca nemestrina | mne-miR-107 | MIMAT0002463 | |
| Monodelphis domestica | mdo-miR-107 | MIMAT0004100 | |
| Ophiophagus hannah | oha-miR-107-3p | MIMAT0036655 | |
| Pan paniscus | ppa-miR-107 | MIMAT0002464 | |
| Pan troglodytes | ptr-miR-107 | MIMAT0002459 | |
| Pig | ssc-miR-107 | MIMAT0002155 | |
| Pongo pygmaeus | ppy-miR-107 | MIMAT0002461 | |
| Rabbit | ocu-miR-107-3p | MIMAT0048200 | |
| Rhesus monkey | mml-miR-107-3p | MIMAT0002458 | |
| Taeniopygia guttata | tgu-miR-107 | MIMAT0014556 | |
| Tetraodon nigroviridis | tni-miR-107 | MIMAT0003044 | |
| Xenopus tropicalis | xtr-miR-107 | MIMAT0003584 | |
| Zebrafish | dre-miR-107a-3p | MIMAT0001817 | |