Search
Search

| Assay Name: | hsa-miR-433-5p |
|---|---|
| Stem-loop Accession Number: | MI0001723 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACGGUGAGCCUGUCAUUAUUC |
| Species: | Human, Mouse, Rat, Dasypus novemcinctus, Guinea pig, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000177, mir-433 |
Mature miRNA Sequence: | UACGGUGAGCCUGUCAUUAUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-433-5p | MIMAT0026554 |
| Mouse | mmu-miR-433-5p | MIMAT0001419 |
| Rat | rno-miR-433-5p | MIMAT0017192 |
| Dasypus novemcinctus | dno-miR-433-5p | MIMAT0047890 |
| Guinea pig | cpo-miR-433-5p | MIMAT0047253 |
| Rabbit | ocu-miR-433-5p | MIMAT0048449 |
| View More Rows | ||
| Rhesus monkey | mml-miR-433-5p | MIMAT0026875 |