Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM10797
          Assay Name: hsa-miR-433-5p
          miRBase Accession Number: MI0001723
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100881886 - 100881978 [+] on Build GRCh38
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Species: Human, Mouse, Rat, Dasypus novemcinctus, Guinea pig, Rabbit, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM10797
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000177, mir-433

          Mature miRNA Sequence:

          UACGGUGAGCCUGUCAUUAUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-433-5p MIMAT0026554
          Mouse mmu-miR-433-5p MIMAT0001419
          Rat rno-miR-433-5p MIMAT0017192
          Dasypus novemcinctus dno-miR-433-5p MIMAT0047890
          Guinea pig cpo-miR-433-5p MIMAT0047253
          Rabbit ocu-miR-433-5p MIMAT0048449
          View More Rows
          Rhesus monkey mml-miR-433-5p MIMAT0026875

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-433
          Stem-loop Accession # MI0001723
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location Chr. 14 - 100881886 - 100881978 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026554
          miRBase ID: hsa-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: Chr. 14 - 100881886 - 100881978 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-433
          Stem-loop Accession # MI0001525
          Stem-loop Sequence
          UGCCCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGGUAGCGGCACCACACCAUGAAGGCAGCCC
          Chromosome Location Chr. 12 - 109591715 - 109591838 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001419
          miRBase ID: mmu-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: Chr. 12 - 109591715 - 109591838 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-433
          Stem-loop Accession # MI0001724
          Stem-loop Sequence
          CCGGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location Chr. 6 - 133712334 - 133712426 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017192
          miRBase ID: rno-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: Chr. 6 - 133712334 - 133712426 [+] on Build Rnor_6.0

          Dasypus novemcinctus

          Stem-loop ID dno-mir-433
          Stem-loop Accession # MI0039110
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCAUGAUCCUCAUCUUUGAGAAGAAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047890
          miRBase ID: dno-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-433
          Stem-loop Accession # MI0038775
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047253
          miRBase ID: cpo-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-433
          Stem-loop Accession # MI0039406
          Stem-loop Sequence
          UACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCGCUGCUGAGAAGGAUCAUGAUGGGCUCCUCGGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048449
          miRBase ID: ocu-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-433
          Stem-loop Accession # MI0007745
          Stem-loop Sequence
          CCAGGGAGAAGUACGGUGAGCCUGUCAUUAUUCAGAGAGGCUAGAUCCUCUGUGUUGAGAAGGAUCAUGAUGGGCUCCUCGGUGUUCUCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026875
          miRBase ID: mml-miR-433-5p
          Mature miRNA Sequence: UACGGUGAGCCUGUCAUUAUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303744_pri
          TaqMan™ Pri-miRNA Assay : Rn03466640_pri
          TaqMan™ Pri-miRNA Assay : Mm03307482_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10797
          TaqMan™ MicroRNA Assay : 001078
          mirVana® miRNA inhibitor : MH10797
          mirVana® miRNA mimic : MC10797
          TaqMan™ Advanced miRNA Assay : mmu478895_mir
          TaqMan™ Advanced miRNA Assay : rno478895_mir
          TaqMan™ Advanced miRNA Assay : 478895_mir


          miRBase Alias:

          Mouse: mmu-miR-433* (v17)
          Rat: rno-miR-433* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0