Search
Search

| Assay Name: | hsa-miR-222-3p |
|---|---|
| miRBase Accession Number: | MI0000299 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGCUACAUCUGGCUACUGGGU |
| Species: | Human, Rat, Bovine, Dog, Horse, Ornithorhynchus anatinus, Pongo pygmaeus, Rhesus monkey, Sarcophilus harrisii, Tupaia chinensis |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000051, mir-221 |
Mature miRNA Sequence: | AGCUACAUCUGGCUACUGGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-222-3p | MIMAT0000279 |
| Rat | rno-miR-222-3p | MIMAT0000891 |
| Bovine | bta-miR-222 | MIMAT0003530 |
| Dog | cfa-miR-222 | MIMAT0009851 |
| Horse | eca-miR-222 | MIMAT0013204 |
| Ornithorhynchus anatinus | oan-miR-222a-3p | MIMAT0007133 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-222 | MIMAT0015808 |
| Rhesus monkey | mml-miR-222-3p | MIMAT0006250 |
| Sarcophilus harrisii | sha-miR-222 | MIMAT0022796 |
| Tupaia chinensis | tch-miR-222-3p | MIMAT0036600 |