Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH11376
          Assay Name: hsa-miR-222-3p
          miRBase Accession Number: MI0000299
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 45747015 - 45747124 [-] on Build GRCh38
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Species: Human, Rat, Bovine, Dog, Horse, Ornithorhynchus anatinus, Pongo pygmaeus, Rhesus monkey, Sarcophilus harrisii, Tupaia chinensis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH11376

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000051, mir-221

          Mature miRNA Sequence:

          AGCUACAUCUGGCUACUGGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-222-3p MIMAT0000279
          Rat rno-miR-222-3p MIMAT0000891
          Bovine bta-miR-222 MIMAT0003530
          Dog cfa-miR-222 MIMAT0009851
          Horse eca-miR-222 MIMAT0013204
          Ornithorhynchus anatinus oan-miR-222a-3p MIMAT0007133
          View More Rows
          Pongo pygmaeus ppy-miR-222 MIMAT0015808
          Rhesus monkey mml-miR-222-3p MIMAT0006250
          Sarcophilus harrisii sha-miR-222 MIMAT0022796
          Tupaia chinensis tch-miR-222-3p MIMAT0036600

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-222
          Stem-loop Accession # MI0000299
          Stem-loop Sequence
          GCUGCUGGAAGGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUCGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location Chr. X - 45747015 - 45747124 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000279
          miRBase ID: hsa-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: Chr. X - 45747015 - 45747124 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-222
          Stem-loop Accession # MI0000962
          Stem-loop Sequence
          AAGGAUUAGGGUGCCCUCAGUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUGGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCAUCUAGCU
          Chromosome Location Chr. X - 3683919 - 3684021 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000891
          miRBase ID: rno-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: Chr. X - 3683919 - 3684021 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-222
          Stem-loop Accession # MI0004744
          Stem-loop Sequence
          GCUGCUGGAAUGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGUAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003530
          miRBase ID: bta-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-222
          Stem-loop Accession # MI0010346
          Stem-loop Sequence
          CCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGUAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009851
          miRBase ID: cfa-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-222
          Stem-loop Accession # MI0012952
          Stem-loop Sequence
          GCUGCUGGAAAGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013204
          miRBase ID: eca-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-222a
          Stem-loop Accession # MI0006865
          Stem-loop Sequence
          AGAGAGCCUGUAGUUGCCCCUCAAUCGCUCAGUAGUCAGUGUAGAUCCUGUAUUCUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAGGGUACCUUCUAGCUUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007133
          miRBase ID: oan-miR-222a-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-222
          Stem-loop Accession # MI0014873
          Stem-loop Sequence
          GCUGCUGGAAGGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUCGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015808
          miRBase ID: ppy-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-222
          Stem-loop Accession # MI0007680
          Stem-loop Sequence
          GCUGCUGGAAGGUAUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUCGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006250
          miRBase ID: mml-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Sarcophilus harrisii

          Stem-loop ID sha-mir-222
          Stem-loop Accession # MI0019623
          Stem-loop Sequence
          CCUUCCACAGAAUAUCUCCAUGAAAAGAUAUGGUGCCUUCUGCCUGUUUUGUCCCUCAUUCGCUCAGUAGUCAGUGUAGAUCCUGUCCCUUUCAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUAGAGCUUCUCUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022796
          miRBase ID: sha-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-222
          Stem-loop Accession # MI0031267
          Stem-loop Sequence
          CUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGCAGCUACAUCUGGCUACUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036600
          miRBase ID: tch-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303011_pri
          TaqMan™ Pri-miRNA Assay : Rn03465472_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11376
          Ambion® Pre-miR™ miRNA Precursor : PM11376
          TaqMan™ MicroRNA Assay : 002276
          mirVana® miRNA mimic : MC11376
          TaqMan™ Advanced miRNA Assay : 477982_mir
          TaqMan™ Advanced miRNA Assay : rno481305_mir
          TaqMan™ Advanced miRNA Assay : mmu481660_mir


          miRBase Alias:

          Human: hsa-miR-222 (v17)
          Mouse: mmu-miR-222-3p (v21)
          Mouse: mmu-miR-222 (v17)
          Rat: rno-miR-222 (v18)
          Macaca mulatta: mml-miR-222 (v19)
          Ochrobactrum anthropi: oan-miR-222a (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline