Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu481660_mir

          Assay Name:
          Stem-loop Accession Number: MI0000299
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 45747015 - 45747124 [-] on Build GRCh38
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Species: Human, Rat, Bovine, Dog, Horse, Ornithorhynchus anatinus, Pongo pygmaeus, Rhesus monkey, Sarcophilus harrisii, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu481660_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000051, mir-221

          Mature miRNA Sequence:

          AGCUACAUCUGGCUACUGGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-222-3p MIMAT0000279
          Rat rno-miR-222-3p MIMAT0000891
          Bovine bta-miR-222 MIMAT0003530
          Dog cfa-miR-222 MIMAT0009851
          Horse eca-miR-222 MIMAT0013204
          Ornithorhynchus anatinus oan-miR-222a-3p MIMAT0007133
          View More Rows
          Pongo pygmaeus ppy-miR-222 MIMAT0015808
          Rhesus monkey mml-miR-222-3p MIMAT0006250
          Sarcophilus harrisii sha-miR-222 MIMAT0022796
          Tupaia chinensis tch-miR-222-3p MIMAT0036600

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-222
          Stem-loop Accession # MI0000299
          Stem-loop Sequence
          GCUGCUGGAAGGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUCGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location Chr. X - 45747015 - 45747124 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000279
          miRBase ID: hsa-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: Chr. X - 45747015 - 45747124 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-222
          Stem-loop Accession # MI0000962
          Stem-loop Sequence
          AAGGAUUAGGGUGCCCUCAGUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUGGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCAUCUAGCU
          Chromosome Location Chr. X - 3683919 - 3684021 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000891
          miRBase ID: rno-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: Chr. X - 3683919 - 3684021 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-222
          Stem-loop Accession # MI0004744
          Stem-loop Sequence
          GCUGCUGGAAUGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGUAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUACCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003530
          miRBase ID: bta-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-222
          Stem-loop Accession # MI0010346
          Stem-loop Sequence
          CCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGUAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009851
          miRBase ID: cfa-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-222
          Stem-loop Accession # MI0012952
          Stem-loop Sequence
          GCUGCUGGAAAGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013204
          miRBase ID: eca-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-222a
          Stem-loop Accession # MI0006865
          Stem-loop Sequence
          AGAGAGCCUGUAGUUGCCCCUCAAUCGCUCAGUAGUCAGUGUAGAUCCUGUAUUCUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAGGGUACCUUCUAGCUUCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007133
          miRBase ID: oan-miR-222a-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-222
          Stem-loop Accession # MI0014873
          Stem-loop Sequence
          GCUGCUGGAAGGUGUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUCGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015808
          miRBase ID: ppy-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-222
          Stem-loop Accession # MI0007680
          Stem-loop Sequence
          GCUGCUGGAAGGUAUAGGUACCCUCAAUGGCUCAGUAGCCAGUGUAGAUCCUGUCUUUCGUAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUCUAGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006250
          miRBase ID: mml-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Sarcophilus harrisii

          Stem-loop ID sha-mir-222
          Stem-loop Accession # MI0019623
          Stem-loop Sequence
          CCUUCCACAGAAUAUCUCCAUGAAAAGAUAUGGUGCCUUCUGCCUGUUUUGUCCCUCAUUCGCUCAGUAGUCAGUGUAGAUCCUGUCCCUUUCAAUCAGCAGCUACAUCUGGCUACUGGGUCUCUGAUGGCAUCUUAGAGCUUCUCUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022796
          miRBase ID: sha-miR-222
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-222
          Stem-loop Accession # MI0031267
          Stem-loop Sequence
          CUCAGUAGCCAGUGUAGAUCCUGUCUUUUGUAAUCAGCAGCUACAUCUGGCUACUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036600
          miRBase ID: tch-miR-222-3p
          Mature miRNA Sequence: AGCUACAUCUGGCUACUGGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303011_pri
          TaqMan™ Pri-miRNA Assay : Rn03465472_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11376
          Ambion® Pre-miR™ miRNA Precursor : PM11376
          TaqMan™ MicroRNA Assay : 002276
          mirVana® miRNA inhibitor : MH11376
          mirVana® miRNA mimic : MC11376
          TaqMan™ Advanced miRNA Assay : 477982_mir
          TaqMan™ Advanced miRNA Assay : rno481305_mir


          miRBase Alias:

          Mouse: mmu-miR-222-3p (v21)

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline