Search
Search

| Assay Name: | hsa-miR-1302 | ||
|---|---|---|---|
| miRBase Accession Number: | MI0006362 | ||
| miRBase Version: | v22.1 | ||
| Chromosome Location: | Chr.12: 112695034 - 112695176 [-] on Build GRCh38 | ||
| Mature miRNA Sequence: | UUGGGACAUACUUAUGCUAAA | ||
| Species: | Human, Callithrix jacchus, Horse, Pan troglodytes, Pongo pygmaeus | ||
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000456, mir-1302 |
Mature miRNA Sequence: | UUGGGACAUACUUAUGCUAAA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1302 | MIMAT0005890 |
| Callithrix jacchus | cja-miR-1302 | MIMAT0039634 |
| Horse | eca-miR-1302d | MIMAT0012969 |
| Pan troglodytes | ptr-miR-1302 | MIMAT0008021 |
| Pongo pygmaeus | ppy-miR-1302 | MIMAT0016222 |
| Stem-loop ID | hsa-mir-1302-1 |
| Stem-loop Accession # | MI0006362 |
| Stem-loop Sequence | CAGAAAGCCCAGUUAAAUUUGAAUUUCAAGUAAACAAUGAAUAAUUGUGUAUGUAAGAAUAUCCCAUACAAUAUUUGGGACAUACUUAUGCUAAAAAUUAUUCCUUGCUUAUCUGAAAUUCAAAUGUAACUAGGAUUCCUGUA |
| Chromosome Location | Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-10 |
| Stem-loop Accession # | MI0015979 |
| Stem-loop Sequence | GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA |
| Chromosome Location | Chr. 15 - 101960459 - 101960596 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-11 |
| Stem-loop Accession # | MI0015980 |
| Stem-loop Sequence | GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA |
| Chromosome Location | Chr. 19 - 71973 - 72110 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-2 |
| Stem-loop Accession # | MI0006363 |
| Stem-loop Sequence | GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA |
| Chromosome Location | Chr. 1 - 30366 - 30503 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-3 |
| Stem-loop Accession # | MI0006364 |
| Stem-loop Sequence | GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUUUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUUA |
| Chromosome Location | Chr. 2 - 113582959 - 113583096 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-4 |
| Stem-loop Accession # | MI0006365 |
| Stem-loop Sequence | AAUGCAGAAGCACAGCUAAAAUUUGAAUUUCAGAUAAACAAAUUUUUCUUAGAAUAAGUAUGUCUCCAUGCAACAUUUGGGACAUACUUAUGCUAAAAUAUUAUUUGUGUUUCAUCUGAAAUUCAAAUUCAACUGGACAUCCUGUAUUUU |
| Chromosome Location | Chr. 2 - 207269275 - 207269424 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-5 |
| Stem-loop Accession # | MI0006366 |
| Stem-loop Sequence | UGCCCGGCCUCCCAUUAAAUUGGUUUUUCAGACAAAUCACAAAUUUGUUUAGGUAUAAGUAUAUCCCAUGUAAUCUUUGGGACAUACUUAUGCUAAAAUAAUUGUUCCUUGUUGAUUGGAAAUUUUAAUUUUAAUUAGGUGUCCUGUAUU |
| Chromosome Location | Chr. 20 - 50614636 - 50614785 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-6 |
| Stem-loop Accession # | MI0006367 |
| Stem-loop Sequence | AACAAAUAAUUUGGUAAUAUAUGUAUGGCCCACACAAUAUUUAGGACAACAAUAUUUGGGACAUACUUAUGCUAAAAAAGUAUUUGUUGA |
| Chromosome Location | Chr. 7 - 18127220 - 18127309 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-7 |
| Stem-loop Accession # | MI0006368 |
| Stem-loop Sequence | ACAACAUGUUUUUAGGACAUGUAUGUCUGGUGCAAUAAUUGGGACAUACUUAUGCUAAAAAAAUUAGUGUUC |
| Chromosome Location | Chr. 8 - 141786242 - 141786313 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-8 |
| Stem-loop Accession # | MI0006369 |
| Stem-loop Sequence | CCCAUUUAAACUUGAAUUUCAUAUAAACACCGUAAUUUUCAGCAUUAGUGUAUCACAUGCAGUAUUUGGGACAUACUUAUGCUAAAAAAUUAGGUGGUGUUGAUCUGAAAUUCCAGUGUAGAUGGGCA |
| Chromosome Location | Chr. 9 - 97363554 - 97363681 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | hsa-mir-1302-9 |
| Stem-loop Accession # | MI0015978 |
| Stem-loop Sequence | GGAUGCCCAGCUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUUA |
| Chromosome Location | Chr. 9 - 30144 - 30281 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0005890
miRBase ID: hsa-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: Chr. 12 - 112695034 - 112695176 [-] on Build GRCh38 |
| Stem-loop ID | cja-mir-1302 |
| Stem-loop Accession # | MI0032171 |
| Stem-loop Sequence | AAAAUGCAGAAGCACAGCUAAAAUUUGAGUUUCAGAUAAACAAAUUUUUCUUAGAAUAAGUAUAUCUCCAUGCAACAUUUGGGACAUACUUAUGCUAAAACAUUAUUUGUGUUUCAUCUGAAAUUCAAAUUCAACUGGAUAUCCUGUAUUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039634
miRBase ID: cja-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | eca-mir-1302d-4 |
| Stem-loop Accession # | MI0012722 |
| Stem-loop Sequence | UUGGAAUUUAGACAAAUAAUGAAGAAUUUUUUAGUGUAAGUAUGUCUUAUGCAAUAUUUGGGACAUACUUAUGCUAAAAAAUUUAUUAGUUAUCUGAAAUUCAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012969
miRBase ID: eca-miR-1302d Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-1 |
| Stem-loop Accession # | MI0008516 |
| Stem-loop Sequence | CAGAAAGCCCAGUUAAAUUUGAAUUUCAAGUAAACAAUGAAUAAUUGUGUAUGUAAGAAUAUCCCAUACAAUAUUUGGGACAUACUUAUGCUAAAAAUUAUUCCUUGCUUAUCUGAAAUUCAAAUGUAACUAGGAUUCCUGU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-2 |
| Stem-loop Accession # | MI0008517 |
| Stem-loop Sequence | GGAUGCCCAGUUAGUUUGAAUUCUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-3 |
| Stem-loop Accession # | MI0008518 |
| Stem-loop Sequence | GAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-4 |
| Stem-loop Accession # | MI0008519 |
| Stem-loop Sequence | GGAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCGUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-5 |
| Stem-loop Accession # | MI0008520 |
| Stem-loop Sequence | GGAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCUUAGCAUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAGAAUUAAGCAUUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-6 |
| Stem-loop Accession # | MI0008521 |
| Stem-loop Sequence | AACAAAUAAUUUGGUAAUAUAUGUAUGGCCCACACAAUAUUUAGGACAACAAUAUUUGGGACAUACUUAUGCUAAAAAAGUAUUUGUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-7 |
| Stem-loop Accession # | MI0008522 |
| Stem-loop Sequence | ACAACAUGUUUUUAGUACAUGUAUGUCUGGUGCAAUAAUUGGGACAUACUUAUGCUAAAAAAAUUAGUGUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ptr-mir-1302-8 |
| Stem-loop Accession # | MI0008523 |
| Stem-loop Sequence | CCCAUUUAAACUUGAAUUUCAUAUAAACACCGUAAUUUUCAGCAUUAGUGUAUCACAUGCAGUAUUUGGGACAUACUUAUGCUAAAAAAUUAGGUGGUGUUGAUCUGAAAUUCCAGUGUAGAUGGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008021
miRBase ID: ptr-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-1302-1 |
| Stem-loop Accession # | MI0015256 |
| Stem-loop Sequence | GGAUGCCCAGUUAGUUUGAAUUUUAGAUAAACAACGAAUAAUUUCUUAGCCUAAAUAUGUCCCAAGCUUAGUUUGGGACAUACUUAUGCUAAAAAACAUUAUUGGUUGUUUAUCUGAGAUUCAAAAUUAAGCAUUUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0016222
miRBase ID: ppy-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-1302-2 |
| Stem-loop Accession # | MI0015260 |
| Stem-loop Sequence | CCCAUUUAAACUUGAAUUUCAUAUAAACAGAGUAAUUUUCAGCAUUAGUGUAUCACAUGCAGUAUUUGGGACAUACUUAUGCUAAAAAAUUAGGUGGUGUUGAUCUGAAAUUCCAGUGUAGAUGGGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0016222
miRBase ID: ppy-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-1302-3 |
| Stem-loop Accession # | MI0015258 |
| Stem-loop Sequence | AACAAAUAAUUUGGUAAUAUAUGUAUGGCCCACACAAUAUUUAGGACAACAAUAUUUGGGACAUACUUAUGCUAAAAAAGUAUUUGUUGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0016222
miRBase ID: ppy-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |
| Stem-loop ID | ppy-mir-1302-4 |
| Stem-loop Accession # | MI0015259 |
| Stem-loop Sequence | ACAACAUUUUUUUUAGUACAUGUAUGUCUGGUGCAACAAUUGGGACAUACUUAUGCUAAAAAAAUUAGUGUUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0016222
miRBase ID: ppy-miR-1302 Mature miRNA Sequence: UUGGGACAUACUUAUGCUAAA Chromosome Location: NA |