Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480946_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-149-5p
          Stem-loop Accession Number: MI0000478
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 240456001 - 240456089 [+] on Build GRCh38
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Species: Human, Mouse, Rat, Bovine, Dasypus novemcinctus, Dog, Eptesicus fuscus, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480946_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000274, mir-149

          Mature miRNA Sequence:

          UCUGGCUCCGUGUCUUCACUCCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-149-5p MIMAT0000450
          Mouse mmu-miR-149-5p MIMAT0000159
          Rat rno-miR-149-5p MIMAT0035726
          Bovine bta-miR-149-5p MIMAT0024570
          Dasypus novemcinctus dno-miR-149-5p MIMAT0047673
          Dog cfa-miR-149 MIMAT0009884
          View More Rows
          Eptesicus fuscus efu-miR-149 MIMAT0035021
          Gorilla gorilla ggo-miR-149 MIMAT0024144
          Guinea pig cpo-miR-149-5p MIMAT0047026
          Horse eca-miR-149 MIMAT0012973
          Pan troglodytes ptr-miR-149 MIMAT0008043
          Pig ssc-miR-149 MIMAT0018379
          Pongo pygmaeus ppy-miR-149 MIMAT0015773
          Rhesus monkey mml-miR-149-5p MIMAT0006210

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-149
          Stem-loop Accession # MI0000478
          Stem-loop Sequence
          GCCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGCUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGCUGGA
          Chromosome Location Chr. 2 - 240456001 - 240456089 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000450
          miRBase ID: hsa-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: Chr. 2 - 240456001 - 240456089 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-149
          Stem-loop Accession # MI0000171
          Stem-loop Sequence
          GGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCGAGGAGGGAGGGAGGGACGGGGGCGGUGCU
          Chromosome Location Chr. 1 - 92850378 - 92850443 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000159
          miRBase ID: mmu-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: Chr. 1 - 92850378 - 92850443 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-149
          Stem-loop Accession # MI0030351
          Stem-loop Sequence
          GCCGGUGCCCGGGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCGAGGAGGGAGGGAGGGACGGGGGCGGUGCUGGGGCAACUGGA
          Chromosome Location Chr. 9 - 100016441 - 100016529 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0035726
          miRBase ID: rno-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: Chr. 9 - 100016441 - 100016529 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-149
          Stem-loop Accession # MI0021115
          Stem-loop Sequence
          GCCGGCGCCCAGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCACCGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCGGCCGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024570
          miRBase ID: bta-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-149
          Stem-loop Accession # MI0038999
          Stem-loop Sequence
          UCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047673
          miRBase ID: dno-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-149
          Stem-loop Accession # MI0010383
          Stem-loop Sequence
          GCCGGCGCCCGGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009884
          miRBase ID: cfa-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-149
          Stem-loop Accession # MI0028706
          Stem-loop Sequence
          GUGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCUGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035021
          miRBase ID: efu-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-149
          Stem-loop Accession # MI0020691
          Stem-loop Sequence
          UCCCAGGCCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGCUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCGGCCGGAACAACGCGGGUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024144
          miRBase ID: ggo-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-149
          Stem-loop Accession # MI0038658
          Stem-loop Sequence
          UCUGGCUCCGUGUCUUCACUCCCGUGUCUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047026
          miRBase ID: cpo-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-149
          Stem-loop Accession # MI0012725
          Stem-loop Sequence
          GCCGGCGCCCAGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCUGCCGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012973
          miRBase ID: eca-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-149
          Stem-loop Accession # MI0008549
          Stem-loop Sequence
          CCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGCUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008043
          miRBase ID: ptr-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-149
          Stem-loop Accession # MI0016620
          Stem-loop Sequence
          GCCCAGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018379
          miRBase ID: ssc-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-149
          Stem-loop Accession # MI0014839
          Stem-loop Sequence
          GCCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCAAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015773
          miRBase ID: ppy-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-149
          Stem-loop Accession # MI0007640
          Stem-loop Sequence
          GCCGGCGCCCAAGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGCCGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006210
          miRBase ID: mml-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306361_pri
          TaqMan™ Pri-miRNA Assay : Hs03303264_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12788
          Ambion® Pre-miR™ miRNA Precursor : PM12788
          TaqMan™ MicroRNA Assay : 002255
          mirVana® miRNA inhibitor : MH12788
          mirVana® miRNA mimic : MC12788
          TaqMan™ Advanced miRNA Assay : 477917_mir
          TaqMan™ Advanced miRNA Assay : rno480946_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline