Search
Search

| Assay Name: | hsa-miR-149-5p |
|---|---|
| miRBase Accession Number: | MI0000478 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCUGGCUCCGUGUCUUCACUCCC |
| Species: | Human, Mouse, Rat, Bovine, Dasypus novemcinctus, Dog, Eptesicus fuscus, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000274, mir-149 |
Mature miRNA Sequence: | UCUGGCUCCGUGUCUUCACUCCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-149-5p | MIMAT0000450 |
| Mouse | mmu-miR-149-5p | MIMAT0000159 |
| Rat | rno-miR-149-5p | MIMAT0035726 |
| Bovine | bta-miR-149-5p | MIMAT0024570 |
| Dasypus novemcinctus | dno-miR-149-5p | MIMAT0047673 |
| Dog | cfa-miR-149 | MIMAT0009884 |
| View More Rows | ||
| Eptesicus fuscus | efu-miR-149 | MIMAT0035021 |
| Gorilla gorilla | ggo-miR-149 | MIMAT0024144 |
| Guinea pig | cpo-miR-149-5p | MIMAT0047026 |
| Horse | eca-miR-149 | MIMAT0012973 |
| Pan troglodytes | ptr-miR-149 | MIMAT0008043 |
| Pig | ssc-miR-149 | MIMAT0018379 |
| Pongo pygmaeus | ppy-miR-149 | MIMAT0015773 |
| Rhesus monkey | mml-miR-149-5p | MIMAT0006210 |