Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12788
          Assay Name: hsa-miR-149-5p
          miRBase Accession Number: MI0000478
          miRBase Version: v22.1
          Chromosome Location: Chr.2: 240456001 - 240456089 [+] on Build GRCh38
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Species: Human, Mouse, Rat, Bovine, Dasypus novemcinctus, Dog, Eptesicus fuscus, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12788

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000274, mir-149

          Mature miRNA Sequence:

          UCUGGCUCCGUGUCUUCACUCCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-149-5p MIMAT0000450
          Mouse mmu-miR-149-5p MIMAT0000159
          Rat rno-miR-149-5p MIMAT0035726
          Bovine bta-miR-149-5p MIMAT0024570
          Dasypus novemcinctus dno-miR-149-5p MIMAT0047673
          Dog cfa-miR-149 MIMAT0009884
          View More Rows
          Eptesicus fuscus efu-miR-149 MIMAT0035021
          Gorilla gorilla ggo-miR-149 MIMAT0024144
          Guinea pig cpo-miR-149-5p MIMAT0047026
          Horse eca-miR-149 MIMAT0012973
          Pan troglodytes ptr-miR-149 MIMAT0008043
          Pig ssc-miR-149 MIMAT0018379
          Pongo pygmaeus ppy-miR-149 MIMAT0015773
          Rhesus monkey mml-miR-149-5p MIMAT0006210

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-149
          Stem-loop Accession # MI0000478
          Stem-loop Sequence
          GCCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGCUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGCUGGA
          Chromosome Location Chr. 2 - 240456001 - 240456089 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000450
          miRBase ID: hsa-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: Chr. 2 - 240456001 - 240456089 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-149
          Stem-loop Accession # MI0000171
          Stem-loop Sequence
          GGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCGAGGAGGGAGGGAGGGACGGGGGCGGUGCU
          Chromosome Location Chr. 1 - 92850378 - 92850443 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000159
          miRBase ID: mmu-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: Chr. 1 - 92850378 - 92850443 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-149
          Stem-loop Accession # MI0030351
          Stem-loop Sequence
          GCCGGUGCCCGGGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCGAGGAGGGAGGGAGGGACGGGGGCGGUGCUGGGGCAACUGGA
          Chromosome Location Chr. 9 - 100016441 - 100016529 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0035726
          miRBase ID: rno-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: Chr. 9 - 100016441 - 100016529 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-149
          Stem-loop Accession # MI0021115
          Stem-loop Sequence
          GCCGGCGCCCAGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCACCGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCGGCCGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024570
          miRBase ID: bta-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-149
          Stem-loop Accession # MI0038999
          Stem-loop Sequence
          UCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047673
          miRBase ID: dno-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-149
          Stem-loop Accession # MI0010383
          Stem-loop Sequence
          GCCGGCGCCCGGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009884
          miRBase ID: cfa-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-149
          Stem-loop Accession # MI0028706
          Stem-loop Sequence
          GUGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCUGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035021
          miRBase ID: efu-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-149
          Stem-loop Accession # MI0020691
          Stem-loop Sequence
          UCCCAGGCCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGCUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCGGCCGGAACAACGCGGGUCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024144
          miRBase ID: ggo-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-149
          Stem-loop Accession # MI0038658
          Stem-loop Sequence
          UCUGGCUCCGUGUCUUCACUCCCGUGUCUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047026
          miRBase ID: cpo-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-149
          Stem-loop Accession # MI0012725
          Stem-loop Sequence
          GCCGGCGCCCAGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCUGCCGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012973
          miRBase ID: eca-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-149
          Stem-loop Accession # MI0008549
          Stem-loop Sequence
          CCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGCUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008043
          miRBase ID: ptr-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-149
          Stem-loop Accession # MI0016620
          Stem-loop Sequence
          GCCCAGGCUCUGGCUCCGUGUCUUCACUCCCGUGUGUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0018379
          miRBase ID: ssc-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-149
          Stem-loop Accession # MI0014839
          Stem-loop Sequence
          GCCGGCGCCCGAGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCAAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015773
          miRBase ID: ppy-miR-149
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-149
          Stem-loop Accession # MI0007640
          Stem-loop Sequence
          GCCGGCGCCCAAGCUCUGGCUCCGUGUCUUCACUCCCGUGUUUGUCCGAGGAGGGAGGGAGGGACGGGGGCUGUGCUGGGGCAGCCGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006210
          miRBase ID: mml-miR-149-5p
          Mature miRNA Sequence: UCUGGCUCCGUGUCUUCACUCCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306361_pri
          TaqMan™ Pri-miRNA Assay : Hs03303264_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12788
          Ambion® Pre-miR™ miRNA Precursor : PM12788
          TaqMan™ MicroRNA Assay : 002255
          mirVana® miRNA inhibitor : MH12788
          TaqMan™ Advanced miRNA Assay : 477917_mir
          TaqMan™ Advanced miRNA Assay : rno480946_mir
          TaqMan™ Advanced miRNA Assay : mmu480946_mir


          miRBase Alias:

          Human: hsa-miR-149 (v17)
          Mouse: mmu-miR-149 (v17)
          Macaca mulatta: mml-miR-149 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline