Search
Search

| Assay Name: | ath-miR395b |
|---|---|
| miRBase Accession Number: | MI0001008 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGAAGUGUUUGGGGGGACUC |
| Species: | Arabidopsis, Arabidopsis lyrata, Brassica napus, Camelina sativa |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000016, MIR395 |
Mature miRNA Sequence: | CUGAAGUGUUUGGGGGGACUC |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Arabidopsis | ath-miR395b | MIMAT0000939 | |
| Arabidopsis | ath-miR395c | MIMAT0000940 | |
| Arabidopsis | ath-miR395f | MIMAT0000943 | |
| Arabidopsis lyrata | aly-miR395b-3p | MIMAT0017548 | |
| Arabidopsis lyrata | aly-miR395f-3p | MIMAT0017556 | |
| Arabidopsis lyrata | aly-miR395h-3p | MIMAT0017668 | |
| View More Rows | |||
| Brassica napus | bna-miR395d | MIMAT0023639 | |
| Brassica napus | bna-miR395e | MIMAT0023640 | |
| Brassica napus | bna-miR395f | MIMAT0023641 | |
| Camelina sativa | cas-miR395b | MIMAT0045341 | |
| Camelina sativa | cas-miR395c-3p | MIMAT0045343 | |