Search
Search

| Assay Name: | ath-miR395b |
|---|---|
| miRBase Accession Number: | MI0001008 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGAAGUGUUUGGGGGGACUC |
| Species: | Arabidopsis, Arabidopsis lyrata, Brassica napus, Camelina sativa |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000016, MIR395 |
Mature miRNA Sequence: | CUGAAGUGUUUGGGGGGACUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Arabidopsis | ath-miR395b | MIMAT0000939 |
| Arabidopsis | ath-miR395c | MIMAT0000940 |
| Arabidopsis | ath-miR395f | MIMAT0000943 |
| Arabidopsis lyrata | aly-miR395b-3p | MIMAT0017548 |
| Arabidopsis lyrata | aly-miR395f-3p | MIMAT0017556 |
| Arabidopsis lyrata | aly-miR395h-3p | MIMAT0017668 |
| View More Rows | ||
| Brassica napus | bna-miR395d | MIMAT0023639 |
| Brassica napus | bna-miR395e | MIMAT0023640 |
| Brassica napus | bna-miR395f | MIMAT0023641 |
| Camelina sativa | cas-miR395b | MIMAT0045341 |
| Camelina sativa | cas-miR395c-3p | MIMAT0045343 |