Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10620
          Assay Name: ath-miR395b
          miRBase Accession Number: MI0001008
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 9364471 - 9364570 [+] on Build TAIR10.1
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Species: Arabidopsis, Arabidopsis lyrata, Brassica napus, Camelina sativa
          Product Type: mirVana® miRNA mimic
          Assay ID MC10620

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000016, MIR395

          Mature miRNA Sequence:

          CUGAAGUGUUUGGGGGGACUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Arabidopsis ath-miR395b MIMAT0000939
          Arabidopsis ath-miR395c MIMAT0000940
          Arabidopsis ath-miR395f MIMAT0000943
          Arabidopsis lyrata aly-miR395b-3p MIMAT0017548
          Arabidopsis lyrata aly-miR395f-3p MIMAT0017556
          Arabidopsis lyrata aly-miR395h-3p MIMAT0017668
          View More Rows
          Brassica napus bna-miR395d MIMAT0023639
          Brassica napus bna-miR395e MIMAT0023640
          Brassica napus bna-miR395f MIMAT0023641
          Camelina sativa cas-miR395b MIMAT0045341
          Camelina sativa cas-miR395c-3p MIMAT0045343

          Stem-loop Details



          Arabidopsis

          Stem-loop ID ath-MIR395b
          Stem-loop Accession # MI0001008
          Stem-loop Sequence
          AUGUCCCCAUGAGUUCCCUUUAACGCUUCAUUGUUAAAUACUCAAAGCCACAUUGGUUUGUAUACAACACUGAAGUGUUUGGGGGGACUCUUGGUGUCAU
          Chromosome Location Chr. 1 - 9364471 - 9364570 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000939
          miRBase ID: ath-miR395b
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: Chr. 1 - 9364471 - 9364570 [+] on Build TAIR10.1

          Stem-loop ID ath-MIR395c
          Stem-loop Accession # MI0001009
          Stem-loop Sequence
          AUGUCCACAUGAGUUCCCUUUAACGCUUCAUUGUUGAAUACUCAAAGCCACAUUGGUUUGUAUAUAACACUGAAGUGUUUGGGGGGACUCUUGGUGUCAU
          Chromosome Location Chr. 1 - 9367080 - 9367179 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000940
          miRBase ID: ath-miR395c
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: Chr. 1 - 9367080 - 9367179 [+] on Build TAIR10.1

          Stem-loop ID ath-MIR395f
          Stem-loop Accession # MI0001012
          Stem-loop Sequence
          AUGUCCCCUUGAGUUCCCUUAAACGCUUCAUUGUUCAUACUUUGUUAUCAUCUAUCGAUCGAUCAAUCAAUCUGAUGAACACUGAAGUGUUUGGGGGGACUCUAGGUGACAU
          Chromosome Location Chr. 1 - 26273858 - 26273969 [+] on Build TAIR10.1
          Mature MicroRNA miRBase Accession #: MIMAT0000943
          miRBase ID: ath-miR395f
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: Chr. 1 - 26273858 - 26273969 [+] on Build TAIR10.1

          Arabidopsis lyrata

          Stem-loop ID aly-MIR395b
          Stem-loop Accession # MI0014576
          Stem-loop Sequence
          GCCAAAUGUCCCCAUGAGUUCCCUUUAACGCUUCAUUGUUAAAUACUCAAAGCCACAUUGGUUUGUAUAUAACACUGAAGUGUUUGGGGGGACUCUUGGUGCCAUUUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017548
          miRBase ID: aly-miR395b-3p
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Stem-loop ID aly-MIR395f
          Stem-loop Accession # MI0014580
          Stem-loop Sequence
          GUCAAAUGUCUCCUUGAGUUCCCUUAAACGCUUUAUUGUUCACACUUUGUUGUCUAAGAUCAAUCGAUCGAUUCAAUCAAUCUGACGAACACUGAAGUGUUUGGGGGGACUCUAGGUGACAUAUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017556
          miRBase ID: aly-miR395f-3p
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Stem-loop ID aly-MIR395h
          Stem-loop Accession # MI0014635
          Stem-loop Sequence
          GUCAACUGUCCCCUUGAGUUCCCUUAAACGCUUCAUUGUUCAUACUUUGUGAGUUUGUGUUCUAAGAUCUAUCGACCGAUAAAUCAAUCUGAUGAACACUGAAGUGUUUGGGGGGACUCUAGGUGACAUCUUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017668
          miRBase ID: aly-miR395h-3p
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Brassica napus

          Stem-loop ID bna-MIR395d
          Stem-loop Accession # MI0020286
          Stem-loop Sequence
          GCCCCCAUGAGUUCCCUUUAACGCUUCAUUGUGAUUACUCCAGUCACAUUUAUUUUUCUUAAAUGUUUGGUCGAUUUUUGUAUACAACACUGAAGUGUUUGGGGGGACUCGUGGUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023639
          miRBase ID: bna-miR395d
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Stem-loop ID bna-MIR395e
          Stem-loop Accession # MI0020287
          Stem-loop Sequence
          CCCCUUGAGUUCCCUGAAACGCUUCAUUGUUCAUUACGUAGAUGUUUAGUAAAUCAAUCUAAUGAACACUGAAGUGUUUGGGGGGACUCUAGGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023640
          miRBase ID: bna-miR395e
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Stem-loop ID bna-MIR395f
          Stem-loop Accession # MI0020288
          Stem-loop Sequence
          GUCCUCUUGAGUUCCCUUUAACGCUUCAUUGUUCAGACUUGGUUGUCUAAUAUCUACCGAUACAUCAACGUAAUGAACACUGAAGUGUUUGGGGGGACUCUAGGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023641
          miRBase ID: bna-miR395f
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Camelina sativa

          Stem-loop ID cas-MIR395b
          Stem-loop Accession # MI0037221
          Stem-loop Sequence
          CCUUAAGUUCCCUUAAACGCUUCACUGUUCAUACUUUGUUUUUAAGAUCUAUCGAUGGAUAAAUUAAUCUGAUGAACACUGAAGUGUUUGGGGGGACUCUAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045341
          miRBase ID: cas-miR395b
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA

          Stem-loop ID cas-MIR395c
          Stem-loop Accession # MI0037222
          Stem-loop Sequence
          CCCAUGAGUUCCCUUUAACGCUUCAUUGUUGUUAAUACACCAAAAACACAUUUGUUUUAUUAAAAUGUUUGUUCGGUUUGUAUAUAACAACACUGAAGUGUUUGGGGGGACUCUUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0045343
          miRBase ID: cas-miR395c-3p
          Mature miRNA Sequence: CUGAAGUGUUUGGGGGGACUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 000365
          mirVana® miRNA inhibitor : MH10620


          miRBase Alias:

          Arabidopsis lyrata: aly-miR395h (v18)
          Arabidopsis lyrata: aly-miR395b (v18)
          Arabidopsis lyrata: aly-miR395f (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline