Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000417
          Assay Name: hsa-miR-30a-5p
          miRBase Accession Number: MI0000088
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 71403551 - 71403621 [-] on Build GRCh38
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Species: Human, Mouse, Rat, Anolis carolinensis, Chicken, Gorilla gorilla, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Xenopus tropicalis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000417
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          UGUAAACAUCCUCGACUGGAAG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30a-5p MIMAT0000087
          Mouse mmu-miR-30a-5p MIMAT0000128
          Rat rno-miR-30a-5p MIMAT0000808
          Anolis carolinensis aca-miR-30a-5p MIMAT0021920
          Chicken gga-miR-30a-5p MIMAT0001135
          Gorilla gorilla ggo-miR-30a-5p MIMAT0002373
          View More Rows
          Monodelphis domestica mdo-miR-30a-5p MIMAT0004093
          Pan paniscus ppa-miR-30a-5p MIMAT0002377
          Pan troglodytes ptr-miR-30a-5p MIMAT0002371
          Pig ssc-miR-30a-5p MIMAT0010193
          Pongo pygmaeus ppy-miR-30a-5p MIMAT0002375
          Rhesus monkey mml-miR-30a-5p MIMAT0002369
          Xenopus tropicalis xtr-miR-30a-5p MIMAT0003574

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30a
          Stem-loop Accession # MI0000088
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location Chr. 6 - 71403551 - 71403621 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000087
          miRBase ID: hsa-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: Chr. 6 - 71403551 - 71403621 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-30a
          Stem-loop Accession # MI0000144
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAAAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location Chr. 1 - 23272269 - 23272339 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000128
          miRBase ID: mmu-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: Chr. 1 - 23272269 - 23272339 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-30a
          Stem-loop Accession # MI0000870
          Stem-loop Sequence
          GCAACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAAAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location Chr. 9 - 29542635 - 29542705 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000808
          miRBase ID: rno-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: Chr. 9 - 29542635 - 29542705 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-30a
          Stem-loop Accession # MI0018847
          Stem-loop Sequence
          UGUUGACGGUGAGCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGUAGAUGAAGCUUUCAGUCGGAUGUUUGCAGCUGCCAACUGCCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021920
          miRBase ID: aca-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-30a
          Stem-loop Accession # MI0001204
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001135
          miRBase ID: gga-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-30a
          Stem-loop Accession # MI0002668
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002373
          miRBase ID: ggo-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-30a
          Stem-loop Accession # MI0005277
          Stem-loop Sequence
          GCGGCUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGCAGAUGGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004093
          miRBase ID: mdo-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-30a
          Stem-loop Accession # MI0002670
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002377
          miRBase ID: ppa-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-30a
          Stem-loop Accession # MI0002667
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002371
          miRBase ID: ptr-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-30a
          Stem-loop Accession # MI0010688
          Stem-loop Sequence
          UCCCUGCGACAGUGAGCGGCUGUAAACAUCCUCGACUGGAAGCUGUGAGGCUGAAGACGGGCUUUCAGUCGGAUGUUUGCAGCCGCCGACUGCCUCCCGCAUCAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0010193
          miRBase ID: ssc-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-30a
          Stem-loop Accession # MI0002669
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCACAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002375
          miRBase ID: ppy-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-30a
          Stem-loop Accession # MI0002666
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCCAUAGAUGGGCUUUCAGUCGGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002369
          miRBase ID: mml-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-30a
          Stem-loop Accession # MI0004813
          Stem-loop Sequence
          GCGACUGUAAACAUCCUCGACUGGAAGCUGUGAAGCAGUUGAAGGGCUUUCAGUCAGAUGUUUGCAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003574
          miRBase ID: xtr-miR-30a-5p
          Mature miRNA Sequence: UGUAAACAUCCUCGACUGGAAG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Human Pool B v3.0
          Megaplex™ RT Primers, Rodent Pool A


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Human MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Human MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306197_pri
          TaqMan™ Pri-miRNA Assay : Rn03465253_pri
          TaqMan™ Pri-miRNA Assay : Hs03302677_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11062
          Ambion® Pre-miR™ miRNA Precursor : PM11062
          mirVana® miRNA inhibitor : MH11062
          mirVana® miRNA mimic : MC11062
          TaqMan™ Advanced miRNA Assay : mmu479448_mir
          TaqMan™ Advanced miRNA Assay : rno479448_mir
          TaqMan™ Advanced miRNA Assay : 479448_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v2.0
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          miRNA Card : TaqMan® Human MicroRNA B Cards v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool B v3.0
          MIRNA : TaqMan® Array Human MicroRNA B Cards v3.0
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          FGMIR : TaqManTM Human MicroRNA Array B v3
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0


          miRBase Alias:

          Human: hsa-miR-30a (v17)
          Mouse: mmu-miR-30a (v17)
          Rat: rno-miR-30a (v18)
          Aeromonas caviae Ae398: aca-miR-30a (v18)
          Monodelphis domestica: mdo-miR-30a (v19)
          Staphylococcus schleiferi: ssc-miR-30a (v14)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline