Search
Search

| Assay Name: | hsa-miR-30a-5p |
|---|---|
| miRBase Accession Number: | MI0000088 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUAAACAUCCUCGACUGGAAG |
| Species: | Human, Mouse, Rat, Anolis carolinensis, Chicken, Gorilla gorilla, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000005, mir-30 |
Mature miRNA Sequence: | UGUAAACAUCCUCGACUGGAAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-30a-5p | MIMAT0000087 |
| Mouse | mmu-miR-30a-5p | MIMAT0000128 |
| Rat | rno-miR-30a-5p | MIMAT0000808 |
| Anolis carolinensis | aca-miR-30a-5p | MIMAT0021920 |
| Chicken | gga-miR-30a-5p | MIMAT0001135 |
| Gorilla gorilla | ggo-miR-30a-5p | MIMAT0002373 |
| View More Rows | ||
| Monodelphis domestica | mdo-miR-30a-5p | MIMAT0004093 |
| Pan paniscus | ppa-miR-30a-5p | MIMAT0002377 |
| Pan troglodytes | ptr-miR-30a-5p | MIMAT0002371 |
| Pig | ssc-miR-30a-5p | MIMAT0010193 |
| Pongo pygmaeus | ppy-miR-30a-5p | MIMAT0002375 |
| Rhesus monkey | mml-miR-30a-5p | MIMAT0002369 |
| Xenopus tropicalis | xtr-miR-30a-5p | MIMAT0003574 |