Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 000517
          Assay Name: hsa-miR-214
          miRBase Accession Number: MI0000954
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 80130905 - 80131014 [+] on Build Rnor_6.0
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Species: Rat, Alligator mississippiensis, Ateles geoffroyi, Chicken, Fugu rubripes, Gadus morhua, Gorilla gorilla, Macaca nemestrina, Monodelphis domestica, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey, Saguinus labiatus, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 000517
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000062, mir-214

          Mature miRNA Sequence:

          ACAGCAGGCACAGACAGGCAG

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-214-3p MIMAT0000885
          Alligator mississippiensis ami-miR-214-3p MIMAT0038239
          Ateles geoffroyi age-miR-214 MIMAT0002551
          Chicken gga-miR-214 MIMAT0007751
          Fugu rubripes fru-miR-214 MIMAT0003026
          Gadus morhua gmo-miR-214-3p MIMAT0044209
          View More Rows
          Gorilla gorilla ggo-miR-214 MIMAT0002550
          Macaca nemestrina mne-miR-214 MIMAT0002557
          Monodelphis domestica mdo-miR-214-3p MIMAT0004130
          Pan paniscus ppa-miR-214 MIMAT0002552
          Pan troglodytes ptr-miR-214 MIMAT0002554
          Pig ssc-miR-214-3p MIMAT0002147
          Pongo pygmaeus ppy-miR-214 MIMAT0002553
          Rhesus monkey mml-miR-214-3p MIMAT0002555
          Saguinus labiatus sla-miR-214 MIMAT0002556
          Tetraodon nigroviridis tni-miR-214 MIMAT0003027
          Xenopus laevis xla-miR-214-3p MIMAT0046535
          Xenopus tropicalis xtr-miR-214 MIMAT0003627
          Zebrafish dre-miR-214 MIMAT0001283

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-214
          Stem-loop Accession # MI0000954
          Stem-loop Sequence
          GUCCUGGAUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location Chr. 13 - 80130905 - 80131014 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000885
          miRBase ID: rno-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: Chr. 13 - 80130905 - 80131014 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-214
          Stem-loop Accession # MI0029797
          Stem-loop Sequence
          CGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCACUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038239
          miRBase ID: ami-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-214
          Stem-loop Accession # MI0002854
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002551
          miRBase ID: age-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-214
          Stem-loop Accession # MI0008208
          Stem-loop Sequence
          GGACUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007751
          miRBase ID: gga-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-214
          Stem-loop Accession # MI0003356
          Stem-loop Sequence
          CAGGAUGAGACUGUUGUGGUGUGUCUGCCUGUCUACACUUGCUGUGCAGACCUUCUGCUCCUGUACAGCAGGCACAGACAGGCAGACAGAUGGCAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003026
          miRBase ID: fru-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-214-1
          Stem-loop Accession # MI0036126
          Stem-loop Sequence
          UGCCUGUCUACACUUGCUGUGCAGACAUUCUGCUCCUGUACAGCAGGCACAGACAGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044209
          miRBase ID: gmo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Stem-loop ID gmo-mir-214-2
          Stem-loop Accession # MI0037985
          Stem-loop Sequence
          UGCCUGUCUUCACUUGCUGUGCAGUAAUUGGUUCAGCCUGUACAGCAGGCACAGACAGGCAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044209
          miRBase ID: gmo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-214
          Stem-loop Accession # MI0002853
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002550
          miRBase ID: ggo-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-214
          Stem-loop Accession # MI0002860
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002557
          miRBase ID: mne-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-214
          Stem-loop Accession # MI0005319
          Stem-loop Sequence
          GGGCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004130
          miRBase ID: mdo-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-214
          Stem-loop Accession # MI0002855
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002552
          miRBase ID: ppa-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-214
          Stem-loop Accession # MI0002857
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002554
          miRBase ID: ptr-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-214
          Stem-loop Accession # MI0002441
          Stem-loop Sequence
          GGCCUGGCUGGACGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002147
          miRBase ID: ssc-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-214
          Stem-loop Accession # MI0002856
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002553
          miRBase ID: ppy-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-214
          Stem-loop Accession # MI0002858
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002555
          miRBase ID: mml-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-214
          Stem-loop Accession # MI0002859
          Stem-loop Sequence
          GGCCUGGCUGGACAGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCGCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002556
          miRBase ID: sla-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-214
          Stem-loop Accession # MI0003357
          Stem-loop Sequence
          GUGGUGUGUCUGCCUGUCUACACUUGCUGUGCAGACCUUCUGCUCCUGUACAGCAGGCACAGACAGGCAGACAGAUGGCAGCCCCACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003027
          miRBase ID: tni-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-214-1
          Stem-loop Accession # MI0038296
          Stem-loop Sequence
          GCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046535
          miRBase ID: xla-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Stem-loop ID xla-mir-214-2
          Stem-loop Accession # MI0038297
          Stem-loop Sequence
          GCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046535
          miRBase ID: xla-miR-214-3p
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-214
          Stem-loop Accession # MI0004867
          Stem-loop Sequence
          UUGGCUGGAAGGAGUUGUCAUGUGUCUGCCUGUCUACACUUGCUGUGCAGAACAUCCUCUCACCUGUACAGCAGGCACAGACAGGCAGUCACAUGACAACCCAGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003627
          miRBase ID: xtr-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-214
          Stem-loop Accession # MI0001381
          Stem-loop Sequence
          UGACUGAGAGCGUUGUCUGUCUGCCUGUCUACACUUGCUGUGCAGAACUUCCUGCACCUGUACAGCAGGCACAGACAGGCAGACAGAUGGCAGCCCGCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001283
          miRBase ID: dre-miR-214
          Mature miRNA Sequence: ACAGCAGGCACAGACAGGCAG
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Rodent MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465830_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10347
          Ambion® Pre-miR™ miRNA Precursor : PM10347
          mirVana® miRNA inhibitor : MH10347
          mirVana® miRNA mimic : MC10347
          TaqMan™ Advanced miRNA Assay : rno480997_mir
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0


          miRBase Alias:

          Human: hsa-miR-214 (v9.2)
          Mouse: mmu-miR-214 (v9.2)
          Rat: rno-miR-214 (v18)
          Macaca mulatta: mml-miR-214 (v19)
          Monodelphis domestica: mdo-miR-214 (v19)
          Staphylococcus schleiferi: ssc-miR-214 (v21)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline