Search
Search

| Assay Name: | hsa-miR-493 |
|---|---|
| miRBase Accession Number: | MI0003132 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGUACAUGGUAGGCUUUCAUU |
| Species: | Human, Rat, Callithrix jacchus, Goat, Pan paniscus, Pig, Pteropus alecto, Rhesus monkey, Sheep |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000230, mir-493 |
Mature miRNA Sequence: | UUGUACAUGGUAGGCUUUCAUU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-493-5p | MIMAT0002813 | |
| Rat | rno-miR-493-5p | MIMAT0017217 | |
| Callithrix jacchus | cja-miR-493 | MIMAT0039530 | |
| Goat | chi-miR-493-5p | MIMAT0036253 | |
| Pan paniscus | ppa-miR-493 | MIMAT0049052 | |
| Pig | ssc-miR-493-5p | MIMAT0025377 | |
| View More Rows | |||
| Pteropus alecto | pal-miR-493-5p | MIMAT0039943 | |
| Rhesus monkey | mml-miR-493-5p | MIMAT0026884 | |
| Sheep | oar-miR-493-5p | MIMAT0019237 | |