Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10627
          Assay Name: hsa-miR-493-5p
          miRBase Accession Number: MI0003132
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100869060 - 100869148 [+] on Build GRCh38
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Species: Human, Rat, Callithrix jacchus, Goat, Pan paniscus, Pig, Pteropus alecto, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA mimic
          Assay ID MC10627

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000230, mir-493

          Mature miRNA Sequence:

          UUGUACAUGGUAGGCUUUCAUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-493-5p MIMAT0002813
          Rat rno-miR-493-5p MIMAT0017217
          Callithrix jacchus cja-miR-493 MIMAT0039530
          Goat chi-miR-493-5p MIMAT0036253
          Pan paniscus ppa-miR-493 MIMAT0049052
          Pig ssc-miR-493-5p MIMAT0025377
          View More Rows
          Pteropus alecto pal-miR-493-5p MIMAT0039943
          Rhesus monkey mml-miR-493-5p MIMAT0026884
          Sheep oar-miR-493-5p MIMAT0019237

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-493
          Stem-loop Accession # MI0003132
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location Chr. 14 - 100869060 - 100869148 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002813
          miRBase ID: hsa-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: Chr. 14 - 100869060 - 100869148 [+] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-493
          Stem-loop Accession # MI0003540
          Stem-loop Sequence
          GGCCUCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCC
          Chromosome Location Chr. 6 - 133699509 - 133699592 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017217
          miRBase ID: rno-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: Chr. 6 - 133699509 - 133699592 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-493
          Stem-loop Accession # MI0032060
          Stem-loop Sequence
          GCGCUCAUUCUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGUAUCGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039530
          miRBase ID: cja-miR-493
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-493
          Stem-loop Accession # MI0030807
          Stem-loop Sequence
          GGGGCUCCCUCUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGCAGCAAGUAAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036253
          miRBase ID: chi-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-493
          Stem-loop Accession # MI0039760
          Stem-loop Sequence
          UUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049052
          miRBase ID: ppa-miR-493
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-493
          Stem-loop Accession # MI0022152
          Stem-loop Sequence
          CCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUGCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025377
          miRBase ID: ssc-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-493
          Stem-loop Accession # MI0032412
          Stem-loop Sequence
          UUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039943
          miRBase ID: pal-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-493
          Stem-loop Accession # MI0007767
          Stem-loop Sequence
          CUGGCCUCCAGGGCUUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026884
          miRBase ID: mml-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-493
          Stem-loop Accession # MI0016912
          Stem-loop Sequence
          GGGGCUCCCUCUGGCCCCCAGGGCCUUGUACAUGGUAGGCUUUCAUUCAUUCGUUUGCACAUUCGGUGAAGGUCUACUGUGUGCCAGGCCCUGUGCCAGGCAGCAAGUAAAUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0019237
          miRBase ID: oar-miR-493-5p
          Mature miRNA Sequence: UUGUACAUGGUAGGCUUUCAUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293825_pri
          TaqMan™ Pri-miRNA Assay : Rn03464857_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10627
          Ambion® Pre-miR™ miRNA Precursor : PM10627
          TaqMan™ MicroRNA Assay : 001040
          mirVana® miRNA inhibitor : MH10627
          TaqMan™ Advanced miRNA Assay : 478943_mir
          TaqMan™ Advanced miRNA Assay : rno481401_mir


          miRBase Alias:

          Human: hsa-miR-493* (v17)
          Rat: rno-miR-493* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline