Search
Search

| Assay Name: | hsa-miR-493-5p |
|---|---|
| miRBase Accession Number: | MI0003132 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUGUACAUGGUAGGCUUUCAUU |
| Species: | Human, Rat, Callithrix jacchus, Goat, Pan paniscus, Pig, Pteropus alecto, Rhesus monkey, Sheep |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000230, mir-493 |
Mature miRNA Sequence: | UUGUACAUGGUAGGCUUUCAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-493-5p | MIMAT0002813 |
| Rat | rno-miR-493-5p | MIMAT0017217 |
| Callithrix jacchus | cja-miR-493 | MIMAT0039530 |
| Goat | chi-miR-493-5p | MIMAT0036253 |
| Pan paniscus | ppa-miR-493 | MIMAT0049052 |
| Pig | ssc-miR-493-5p | MIMAT0025377 |
| View More Rows | ||
| Pteropus alecto | pal-miR-493-5p | MIMAT0039943 |
| Rhesus monkey | mml-miR-493-5p | MIMAT0026884 |
| Sheep | oar-miR-493-5p | MIMAT0019237 |