Search
Search

| Assay Name: | mmu-miR-322 |
|---|---|
| miRBase Accession Number: | MI0000589 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAACAUGAAGCGCUGCAACA |
| Species: | Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000164, mir-322 |
Mature miRNA Sequence: | AAACAUGAAGCGCUGCAACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-322-3p | MIMAT0000547 |