Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 001061
          Assay Name: mmu-miR-326
          miRBase Accession Number: MI0000598
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 99552269 - 99552363 [+] on Build GRCm38
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAGU
          Species: Mouse, Rat, Cricetulus griseus
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 001061
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000143, mir-326

          Mature miRNA Sequence:

          CCUCUGGGCCCUUCCUCCAGU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-326-3p MIMAT0000559
          Rat rno-miR-326-3p MIMAT0000560
          Cricetulus griseus cgr-miR-326 MIMAT0023913

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-326
          Stem-loop Accession # MI0000598
          Stem-loop Sequence
          CUCAUCUGUCUGUUGGGCUGGGGGCAGGGCCUUUGUGAAGGCGGGUUAUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGUCCCGAGGCAGAUUUA
          Chromosome Location Chr. 7 - 99552269 - 99552363 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000559
          miRBase ID: mmu-miR-326-3p
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAGU
          Chromosome Location: Chr. 7 - 99552269 - 99552363 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-326
          Stem-loop Accession # MI0000599
          Stem-loop Sequence
          CUCAUCUGUCUGUGGGGCUGGGGGCAGGGCCUUUGUGAAGGCGGGUUAUGCUCAGAUCGCCUCUGGGCCCUUCCUCCAGUCCCGAGGCAGAUUUA
          Chromosome Location Chr. 1 - 164518401 - 164518495 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000560
          miRBase ID: rno-miR-326-3p
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAGU
          Chromosome Location: Chr. 1 - 164518401 - 164518495 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-326
          Stem-loop Accession # MI0020495
          Stem-loop Sequence
          CUAUUGGGCUGGGGGCAGGGCCUUUGUGAAGGCGGGUUAUGCUCGGAUCGCCUCUGGGCCCUUCCUCCAGUCCCGAGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023913
          miRBase ID: cgr-miR-326
          Mature miRNA Sequence: CCUCUGGGCCCUUCCUCCAGU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pool:

          Megaplex™ RT Primers, Rodent Pool B v3.0


          Associated TaqMan® Array MicroRNA Card:

          TaqMan® Rodent MicroRNA B Cards v3.0


          TaqMan® OpenArray® MicroRNA Panel:

          TaqMan® OpenArray® Rodent MicroRNA Panel




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306944_pri
          TaqMan™ Pri-miRNA Assay : Rn03465905_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11036
          Ambion® Pre-miR™ miRNA Precursor : PM11036
          mirVana® miRNA inhibitor : MH11036
          mirVana® miRNA mimic : MC11036
          TaqMan™ Advanced miRNA Assay : mmu481778_mir
          TaqMan™ Advanced miRNA Assay : rno481057_mir
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v2.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          MIRNA : TaqMan® Array Rodent MicroRNA B Cards v3.0
          FGMIR : TaqManTM Array Rodent MicroRNA B Card v3
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool B v3.0
          miRNA Card : TaqMan® Rodent MicroRNA B Cards v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate


          miRBase Alias:

          Mouse: mmu-miR-326 (v17)
          Rat: rno-miR-326 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0