Search
Search

| Assay Name: | mmu-miR-199b |
|---|---|
| miRBase Accession Number: | MI0000714 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCCAGUGUUUAGACUACCUGUUC |
| Species: | Mouse, Cricetulus griseus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000040, mir-199 |
Mature miRNA Sequence: | CCCAGUGUUUAGACUACCUGUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-199b-5p | MIMAT0000672 |
| Cricetulus griseus | cgr-miR-199b | MIMAT0023838 |