Search
Search

| Assay Name: | rno-miR-382* |
|---|---|
| miRBase Accession Number: | MI0000790 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAUCAUUCACGGACAACACUU |
| Species: | Human, Rat, Goat, Gorilla gorilla, Pan paniscus, Sheep |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCAUUCACGGACAACACUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-382-3p | MIMAT0022697 |
| Rat | rno-miR-382-3p | MIMAT0003202 |
| Goat | chi-miR-382-3p | MIMAT0036201 |
| Gorilla gorilla | ggo-miR-382 | MIMAT0049116 |
| Pan paniscus | ppa-miR-382 | MIMAT0049038 |
| Sheep | oar-miR-382-3p | MIMAT0019307 |