Search
Search

| Assay Name: | U87 |
|---|---|
| NCBI Accession Number: | AF272707 |
| Control Sequence: | ACAATGATGACTTATGTTTTTGCCGTTTACCCAGCTGAGGGTTTCTTTGAAGAGAGAATCTTAAGACTGAGC |
| Species: | Rat |
| Product Type: | TaqMan™ microRNA Control Assay |
Alias: | U87 |
Control Sequence: | ACAATGATGACTTATGTTTTTGCCGTTTACCCAGCTGAGGGTTTCTTTGAAGAGAGAATCTTAAGACTGAGC |
| Species | NCBI Accession Number |
|---|---|
| AF272707 |