Search
Search

| Assay Name: | mmu-miR-615 |
|---|---|
| miRBase Accession Number: | MI0003628 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCCUCUU |
| Species: | Human, Mouse, Callithrix jacchus, Cricetulus griseus, Guinea pig, Pan troglodytes, Rhesus monkey |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000342, mir-615 |
Mature miRNA Sequence: | UCCGAGCCUGGGUCUCCCUCUU |
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-615-3p | MIMAT0003283 | |
| Mouse | mmu-miR-615-3p | MIMAT0003783 | |
| Callithrix jacchus | cja-miR-615 | MIMAT0039466 | |
| Cricetulus griseus | cgr-miR-615-3p | MIMAT0023988 | |
| Guinea pig | cpo-miR-615-3p | MIMAT0047354 | |
| Pan troglodytes | ptr-miR-615 | MIMAT0008277 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-615-3p | MIMAT0006466 | |