Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs03304632_pri
Assay Name:
hsa-mir-615
Stem-loop ID:
hsa-mir-615
Mapped to miRBase Version:
v22.1
Chromosome Location:
Chr.12: 54033950 - 54034045 [+] on Build GRCh38
Stem-loop Location:
Chr.12: 54033950 - 54034045 [+] on Build GRCh38
Mature miRNA ID:
hsa-miR-615-5p
hsa-miR-615-3p
Species:
Human
Product Type:
TaqMan
™
Pri-miRNA Assay
Assay ID
Hs03304632_pri
Size
S: 360 rxns; 20X
M: 750 rxns; 20X
L: 2900 rxns; 60X
Availability
Made To Order
Catalog #
4427012
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Amplicon Length:
63
Chromosome Location:
Chr. 12 - 54034247 - 54034247 [+] on 54034247 Build GRCh38
Species:
Human
miRBase Details:
Species
miRBase ID
miRBase Accession Number
Human
hsa-miR-615-5p
MIMAT0004804
Human
hsa-miR-615-3p
MIMAT0003283
Stem-loop Details
Human
Stem-loop ID
hsa-mir-615
Stem-loop Accession #
MI0003628
Stem-loop Sequence
CUCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCCC
Chromosome Location
Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38
Mature MicroRNA
miRBase Accession #: MIMAT0004804
miRBase ID: hsa-miR-615-5p
Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
Chromosome Location: Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38
miRBase Accession #: MIMAT0003283
miRBase ID: hsa-miR-615-3p
Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
Chromosome Location: Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38
Gene Family ID
MIPF0000342,
mir-615
Distance to Amplicon
178 bases
Back To Top
More Information
Related Products
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM12460
Ambion
®
Pre-miR
™
miRNA Precursor :
PM12460
TaqMan
™
MicroRNA Assay :
002353
mir
Vana
®
miRNA inhibitor :
MH12460
mir
Vana
®
miRNA mimic :
MC12460
TaqMan
™
Advanced miRNA Assay :
478176_mir
TaqMan
™
Advanced miRNA Assay :
mmu481220_mir
TaqMan
™
Advanced miRNA Assay :
rno481220_mir
Ambion
®
Anti-miR
™
miRNA Inhibitor :
AM11731
Ambion
®
Pre-miR
™
miRNA Precursor :
PM11731
TaqMan
™
MicroRNA Assay :
001960
mir
Vana
®
miRNA inhibitor :
MH11731
mir
Vana
®
miRNA mimic :
MC11731
TaqMan
™
Advanced miRNA Assay :
478175_mir
TaqMan
™
Advanced miRNA Assay :
mmu481930_mir
Back To Top
Related Products
High Capacity RNA-to-cDNA Kit
SuperScript
®
IV VILO
™
Master Mix
TaqMan
®
Universal PCR Master Mix
TaqMan
®
Fast Advanced Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline