Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12460
          Assay Name: hsa-miR-615-5p
          miRBase Accession Number: MI0003628
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 54033950 - 54034045 [+] on Build GRCh38
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Guinea pig, Horse, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12460

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000342, mir-615

          Mature miRNA Sequence:

          GGGGGUCCCCGGUGCUCGGAUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-615-5p MIMAT0004804
          Mouse mmu-miR-615-5p MIMAT0004837
          Rat rno-miR-615 MIMAT0012835
          Bovine bta-miR-615 MIMAT0009355
          Cricetulus griseus cgr-miR-615-5p MIMAT0023987
          Guinea pig cpo-miR-615-5p MIMAT0047353
          View More Rows
          Horse eca-miR-615-5p MIMAT0012976
          Rhesus monkey mml-miR-615-5p MIMAT0006465

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-615
          Stem-loop Accession # MI0003628
          Stem-loop Sequence
          CUCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCCC
          Chromosome Location Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004804
          miRBase ID: hsa-miR-615-5p
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-615
          Stem-loop Accession # MI0005004
          Stem-loop Sequence
          UCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAUCCC
          Chromosome Location Chr. 15 - 103014910 - 103015001 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004837
          miRBase ID: mmu-miR-615-5p
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: Chr. 15 - 103014910 - 103015001 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-615
          Stem-loop Accession # MI0012597
          Stem-loop Sequence
          UCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAUCCC
          Chromosome Location Chr. 7 - 144629019 - 144629110 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0012835
          miRBase ID: rno-miR-615
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: Chr. 7 - 144629019 - 144629110 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-615
          Stem-loop Accession # MI0009877
          Stem-loop Sequence
          CUCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUCCCCCCACCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009355
          miRBase ID: bta-miR-615
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-615
          Stem-loop Accession # MI0020543
          Stem-loop Sequence
          GGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023987
          miRBase ID: cgr-miR-615-5p
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-615
          Stem-loop Accession # MI0038826
          Stem-loop Sequence
          GGGGGUCCCCGGUGCUCGGAUCUCGAGAGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047353
          miRBase ID: cpo-miR-615-5p
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-615
          Stem-loop Accession # MI0012729
          Stem-loop Sequence
          ACGCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUCCCCCCCUCCUCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012976
          miRBase ID: eca-miR-615-5p
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-615
          Stem-loop Accession # MI0007869
          Stem-loop Sequence
          CUCGGGAGGGGCGGAAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006465
          miRBase ID: mml-miR-615-5p
          Mature miRNA Sequence: GGGGGUCCCCGGUGCUCGGAUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308337_pri
          TaqMan™ Pri-miRNA Assay : Hs03304632_pri
          TaqMan™ Pri-miRNA Assay : Rn04229620_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12460
          Ambion® Pre-miR™ miRNA Precursor : PM12460
          TaqMan™ MicroRNA Assay : 002353
          mirVana® miRNA inhibitor : MH12460
          TaqMan™ Advanced miRNA Assay : 478176_mir
          TaqMan™ Advanced miRNA Assay : mmu481220_mir
          TaqMan™ Advanced miRNA Assay : rno481220_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline