Search
Search

| Assay Name: | hsa-miR-615-5p |
|---|---|
| miRBase Accession Number: | MI0003628 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGGGGUCCCCGGUGCUCGGAUC |
| Species: | Human, Mouse, Rat, Bovine, Cricetulus griseus, Guinea pig, Horse, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000342, mir-615 |
Mature miRNA Sequence: | GGGGGUCCCCGGUGCUCGGAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-615-5p | MIMAT0004804 |
| Mouse | mmu-miR-615-5p | MIMAT0004837 |
| Rat | rno-miR-615 | MIMAT0012835 |
| Bovine | bta-miR-615 | MIMAT0009355 |
| Cricetulus griseus | cgr-miR-615-5p | MIMAT0023987 |
| Guinea pig | cpo-miR-615-5p | MIMAT0047353 |
| View More Rows | ||
| Horse | eca-miR-615-5p | MIMAT0012976 |
| Rhesus monkey | mml-miR-615-5p | MIMAT0006465 |