Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11731
          Assay Name: hsa-miR-615-3p
          miRBase Accession Number: MI0003628
          miRBase Version: v22.1
          Chromosome Location: Chr.12: 54033950 - 54034045 [+] on Build GRCh38
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Species: Human, Mouse, Callithrix jacchus, Cricetulus griseus, Guinea pig, Pan troglodytes, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11731
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000342, mir-615

          Mature miRNA Sequence:

          UCCGAGCCUGGGUCUCCCUCUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-615-3p MIMAT0003283
          Mouse mmu-miR-615-3p MIMAT0003783
          Callithrix jacchus cja-miR-615 MIMAT0039466
          Cricetulus griseus cgr-miR-615-3p MIMAT0023988
          Guinea pig cpo-miR-615-3p MIMAT0047354
          Pan troglodytes ptr-miR-615 MIMAT0008277
          View More Rows
          Rhesus monkey mml-miR-615-3p MIMAT0006466

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-615
          Stem-loop Accession # MI0003628
          Stem-loop Sequence
          CUCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCCC
          Chromosome Location Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003283
          miRBase ID: hsa-miR-615-3p
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: Chr. 12 - 54033950 - 54034045 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-615
          Stem-loop Accession # MI0005004
          Stem-loop Sequence
          UCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAUCCC
          Chromosome Location Chr. 15 - 103014910 - 103015001 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003783
          miRBase ID: mmu-miR-615-3p
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: Chr. 15 - 103014910 - 103015001 [+] on Build GRCm38

          Callithrix jacchus

          Stem-loop ID cja-mir-615
          Stem-loop Accession # MI0031990
          Stem-loop Sequence
          CGCUCGCGGCUCCGGAGGAUUCCAGCGACUCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCCCCUCAGCCCCUCCGGCUGCAGAGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039466
          miRBase ID: cja-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-615
          Stem-loop Accession # MI0020543
          Stem-loop Sequence
          GGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023988
          miRBase ID: cgr-miR-615-3p
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-615
          Stem-loop Accession # MI0038826
          Stem-loop Sequence
          GGGGGUCCCCGGUGCUCGGAUCUCGAGAGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047354
          miRBase ID: cpo-miR-615-3p
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-615
          Stem-loop Accession # MI0008814
          Stem-loop Sequence
          CUCGGGAGGGGCGGGAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008277
          miRBase ID: ptr-miR-615
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-615
          Stem-loop Accession # MI0007869
          Stem-loop Sequence
          CUCGGGAGGGGCGGAAGGGGGGUCCCCGGUGCUCGGAUCUCGAGGGUGCUUAUUGUUCGGUCCGAGCCUGGGUCUCCCUCUUCCCCCCAACCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006466
          miRBase ID: mml-miR-615-3p
          Mature miRNA Sequence: UCCGAGCCUGGGUCUCCCUCUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308337_pri
          TaqMan™ Pri-miRNA Assay : Hs03304632_pri
          Ambion® Pre-miR™ miRNA Precursor : PM11731
          TaqMan™ MicroRNA Assay : 001960
          mirVana® miRNA inhibitor : MH11731
          mirVana® miRNA mimic : MC11731
          TaqMan™ Advanced miRNA Assay : 478175_mir
          TaqMan™ Advanced miRNA Assay : mmu481930_mir


          miRBase Alias:

          Mouse: mmu-miR-615 (v9.2)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline