Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002101
          Assay Name: hsa-miR-296-3p
          miRBase Accession Number: MI0000747
          miRBase Version: v22.1
          Chromosome Location: Chr.20: 58817615 - 58817694 [-] on Build GRCh38
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Species: Human, Mouse, Rat, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002101
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000159, mir-296

          Mature miRNA Sequence:

          GAGGGUUGGGUGGAGGCUCUCC

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-296-3p MIMAT0004679
          Mouse mmu-miR-296-3p MIMAT0004576
          Rat rno-miR-296-3p MIMAT0004742
          Pan troglodytes ptr-miR-296 MIMAT0008079
          Pongo pygmaeus ppy-miR-296-3p MIMAT0015810
          Rhesus monkey mml-miR-296-3p MIMAT0006252

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-296
          Stem-loop Accession # MI0000747
          Stem-loop Sequence
          AGGACCCUUCCAGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUCU
          Chromosome Location Chr. 20 - 58817615 - 58817694 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004679
          miRBase ID: hsa-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: Chr. 20 - 58817615 - 58817694 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-296
          Stem-loop Accession # MI0000394
          Stem-loop Sequence
          GGGCCUUUCUGGAGGGCCCCCCCUCAAUCCUGUUGUGCUCGCUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGUGUCC
          Chromosome Location Chr. 2 - 174267047 - 174267125 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004576
          miRBase ID: mmu-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: Chr. 2 - 174267047 - 174267125 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-296
          Stem-loop Accession # MI0000967
          Stem-loop Sequence
          GGACCUUUCUGGAGGGCCCCCCCUCAAUCCUGUUGUGCUCGCUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGUGUC
          Chromosome Location Chr. 3 - 172357490 - 172357567 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004742
          miRBase ID: rno-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: Chr. 3 - 172357490 - 172357567 [-] on Build Rnor_6.0

          Pan troglodytes

          Stem-loop ID ptr-mir-296
          Stem-loop Accession # MI0008592
          Stem-loop Sequence
          AGGACCCUUCCGGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008079
          miRBase ID: ptr-miR-296
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-296
          Stem-loop Accession # MI0014874
          Stem-loop Sequence
          AGGACCCUUCCGGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015810
          miRBase ID: ppy-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-296
          Stem-loop Accession # MI0007681
          Stem-loop Sequence
          AGGACCCUUCCAGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006252
          miRBase ID: mml-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303414_pri
          TaqMan™ Pri-miRNA Assay : Mm03306611_pri
          TaqMan™ Pri-miRNA Assay : Rn03464656_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12294
          Ambion® Pre-miR™ miRNA Precursor : PM12294
          mirVana® miRNA inhibitor : MH12294
          mirVana® miRNA mimic : MC12294
          TaqMan™ Advanced miRNA Assay : 478790_mir
          TaqMan™ Advanced miRNA Assay : mmu481026_mir
          TaqMan™ Advanced miRNA Assay : rno481026_mir
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0
          FGMIR : TaqManTM Human MicroRNA Array A
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0


          miRBase Alias:

          Rat: rno-miR-296 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline