Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481026_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-296-3p
          Stem-loop Accession Number: MI0000747
          miRBase Version: v22.1
          Chromosome Location: Chr.20: 58817615 - 58817694 [-] on Build GRCh38
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Species: Human, Mouse, Rat, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481026_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000159, mir-296

          Mature miRNA Sequence:

          GAGGGUUGGGUGGAGGCUCUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-296-3p MIMAT0004679
          Mouse mmu-miR-296-3p MIMAT0004576
          Rat rno-miR-296-3p MIMAT0004742
          Pan troglodytes ptr-miR-296 MIMAT0008079
          Pongo pygmaeus ppy-miR-296-3p MIMAT0015810
          Rhesus monkey mml-miR-296-3p MIMAT0006252

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-296
          Stem-loop Accession # MI0000747
          Stem-loop Sequence
          AGGACCCUUCCAGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUCU
          Chromosome Location Chr. 20 - 58817615 - 58817694 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004679
          miRBase ID: hsa-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: Chr. 20 - 58817615 - 58817694 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-296
          Stem-loop Accession # MI0000394
          Stem-loop Sequence
          GGGCCUUUCUGGAGGGCCCCCCCUCAAUCCUGUUGUGCUCGCUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGUGUCC
          Chromosome Location Chr. 2 - 174267047 - 174267125 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004576
          miRBase ID: mmu-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: Chr. 2 - 174267047 - 174267125 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-296
          Stem-loop Accession # MI0000967
          Stem-loop Sequence
          GGACCUUUCUGGAGGGCCCCCCCUCAAUCCUGUUGUGCUCGCUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGUGUC
          Chromosome Location Chr. 3 - 172357490 - 172357567 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004742
          miRBase ID: rno-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: Chr. 3 - 172357490 - 172357567 [-] on Build Rnor_6.0

          Pan troglodytes

          Stem-loop ID ptr-mir-296
          Stem-loop Accession # MI0008592
          Stem-loop Sequence
          AGGACCCUUCCGGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008079
          miRBase ID: ptr-miR-296
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-296
          Stem-loop Accession # MI0014874
          Stem-loop Sequence
          AGGACCCUUCCGGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015810
          miRBase ID: ppy-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-296
          Stem-loop Accession # MI0007681
          Stem-loop Sequence
          AGGACCCUUCCAGAGGGCCCCCCCUCAAUCCUGUUGUGCCUAAUUCAGAGGGUUGGGUGGAGGCUCUCCUGAAGGGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006252
          miRBase ID: mml-miR-296-3p
          Mature miRNA Sequence: GAGGGUUGGGUGGAGGCUCUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303414_pri
          TaqMan™ Pri-miRNA Assay : Mm03306611_pri
          TaqMan™ Pri-miRNA Assay : Rn03464656_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12294
          Ambion® Pre-miR™ miRNA Precursor : PM12294
          TaqMan™ MicroRNA Assay : 002101
          mirVana® miRNA inhibitor : MH12294
          mirVana® miRNA mimic : MC12294
          TaqMan™ Advanced miRNA Assay : 478790_mir
          TaqMan™ Advanced miRNA Assay : mmu481026_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline