Search
Search

| Assay Name: | hsa-miR-296-3p |
|---|---|
| Stem-loop Accession Number: | MI0000747 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GAGGGUUGGGUGGAGGCUCUCC |
| Species: | Human, Mouse, Rat, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000159, mir-296 |
Mature miRNA Sequence: | GAGGGUUGGGUGGAGGCUCUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-296-3p | MIMAT0004679 |
| Mouse | mmu-miR-296-3p | MIMAT0004576 |
| Rat | rno-miR-296-3p | MIMAT0004742 |
| Pan troglodytes | ptr-miR-296 | MIMAT0008079 |
| Pongo pygmaeus | ppy-miR-296-3p | MIMAT0015810 |
| Rhesus monkey | mml-miR-296-3p | MIMAT0006252 |