Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 002269
          Assay Name: hsa-miR-183
          miRBase Accession Number: MI0000273
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 129774905 - 129775014 [-] on Build GRCh38
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Branchiostoma floridae, Callithrix jacchus, Dasypus novemcinctus, Dog, Gadus morhua, Goat, Guinea pig, Horse, Ornithorhynchus anatinus, Pongo pygmaeus, Pteropus alecto, Rabbit, Salmo salar, Taeniopygia guttata, Tupaia chinensis
          Product Type: TaqMan™ MicroRNA Assay
          Assay ID 002269
          Size
          Availability Inventoried
          Catalog # 4427975
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000066, mir-183

          Mature miRNA Sequence:

          UAUGGCACUGGUAGAAUUCACU

          Mature miRNA Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-183-5p MIMAT0000261
          Mouse mmu-miR-183-5p MIMAT0000212
          Rat rno-miR-183-5p MIMAT0000860
          Alligator mississippiensis ami-miR-183-5p MIMAT0038196
          Branchiostoma floridae bfl-miR-183 MIMAT0009487
          Callithrix jacchus cja-miR-183 MIMAT0039448
          View More Rows
          Dasypus novemcinctus dno-miR-183-5p MIMAT0047701
          Dog cfa-miR-183 MIMAT0006621
          Gadus morhua gmo-miR-183-5p MIMAT0044261
          Goat chi-miR-183 MIMAT0036007
          Guinea pig cpo-miR-183-5p MIMAT0047050
          Horse eca-miR-183 MIMAT0012938
          Ornithorhynchus anatinus oan-miR-183 MIMAT0006805
          Pongo pygmaeus ppy-miR-183 MIMAT0015780
          Pteropus alecto pal-miR-183-5p MIMAT0040144
          Rabbit ocu-miR-183-5p MIMAT0048287
          Salmo salar ssa-miR-183-5p MIMAT0032429
          Taeniopygia guttata tgu-miR-183 MIMAT0025393
          Tupaia chinensis tch-miR-183-5p MIMAT0036573

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-183
          Stem-loop Accession # MI0000273
          Stem-loop Sequence
          CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA
          Chromosome Location Chr. 7 - 129774905 - 129775014 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000261
          miRBase ID: hsa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 7 - 129774905 - 129775014 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-183
          Stem-loop Accession # MI0000225
          Stem-loop Sequence
          CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAG
          Chromosome Location Chr. 6 - 30169668 - 30169737 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000212
          miRBase ID: mmu-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 6 - 30169668 - 30169737 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-183
          Stem-loop Accession # MI0000928
          Stem-loop Sequence
          CCAGAGAGUGUGACUCCUGUCCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCGCGA
          Chromosome Location Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000860
          miRBase ID: rno-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-183
          Stem-loop Accession # MI0029768
          Stem-loop Sequence
          ACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACGCUAUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038196
          miRBase ID: ami-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-183
          Stem-loop Accession # MI0010031
          Stem-loop Sequence
          CCACGUGCCCGUUCACGGUCUAUGGCACUGGUAGAAUUCACUGAUCGUAGACACCAGUGAUUCUUCAGUCAGCCAUACACCGUGGCGGGAAACUGAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009487
          miRBase ID: bfl-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-183
          Stem-loop Accession # MI0031971
          Stem-loop Sequence
          UGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039448
          miRBase ID: cja-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-183
          Stem-loop Accession # MI0039014
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047701
          miRBase ID: dno-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-183
          Stem-loop Accession # MI0008017
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006621
          miRBase ID: cfa-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-183
          Stem-loop Accession # MI0036163
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044261
          miRBase ID: gmo-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-183
          Stem-loop Accession # MI0030654
          Stem-loop Sequence
          CCAGAGAGCGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCGCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036007
          miRBase ID: chi-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-183
          Stem-loop Accession # MI0038671
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047050
          miRBase ID: cpo-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-183
          Stem-loop Accession # MI0012692
          Stem-loop Sequence
          CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012938
          miRBase ID: eca-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-183
          Stem-loop Accession # MI0006667
          Stem-loop Sequence
          GGAGGGGGAGACCCUCACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGAGAAAGAACCACAAGACUCUCCCAUCUCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006805
          miRBase ID: oan-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-183
          Stem-loop Accession # MI0014848
          Stem-loop Sequence
          CAGAGCGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015780
          miRBase ID: ppy-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-183
          Stem-loop Accession # MI0032516
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040144
          miRBase ID: pal-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-183
          Stem-loop Accession # MI0039322
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048287
          miRBase ID: ocu-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-183-1
          Stem-loop Accession # MI0026544
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGACGGCACACUAUCAGUGAAUUACCAUAGGGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032429
          miRBase ID: ssa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-183-2
          Stem-loop Accession # MI0026545
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAAGGCACACUAUCAGUGAAUUACCAUUGGGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032429
          miRBase ID: ssa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-183-3
          Stem-loop Accession # MI0026546
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAUAGGGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032429
          miRBase ID: ssa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-183
          Stem-loop Accession # MI0022170
          Stem-loop Sequence
          CUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGCAACCCCGCGGUCAGUGAAUUACCAUAGGGCCAUAAACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025393
          miRBase ID: tgu-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-183
          Stem-loop Accession # MI0031236
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036573
          miRBase ID: tch-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA


          Back To Top

          More Information


          Associated Megaplex™ RT Primer Pools:

          Megaplex™ RT Primers, Rodent Pool A
          Megaplex™ RT Primers, Human Pool A




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464630_pri
          TaqMan™ Pri-miRNA Assay : Hs03302720_pri
          TaqMan™ Pri-miRNA Assay : Mm03306888_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12830
          Ambion® Pre-miR™ miRNA Precursor : PM12830
          mirVana® miRNA inhibitor : MH12830
          mirVana® miRNA mimic : MC12830
          TaqMan™ Advanced miRNA Assay : 477937_mir
          TaqMan™ Advanced miRNA Assay : rno481330_mir
          TaqMan™ Advanced miRNA Assay : mmu482690_mir
          FGMIR : TaqManTM Rodent MicroRNA Array A
          MIRNA : TaqMan® OpenArray Rodent MicroRNA Panel
          miRNA Card : TaqMan® Human MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Rodent Pool A
          MIRNA : TaqMan® OpenArray Human MicroRNA Panel
          miRNA Card : TaqMan® Rodent MicroRNA A Cards v2.0
          MIRNA : TaqMan® Array Rodent MicroRNA A+B Cards Set v3.0
          MIRNA : TaqMan® Array Human MicroRNA A+B Cards Set v3.0
          FGMIR : QS TaqMan OpenArray Hu miRNA Plate
          MIRNA : TaqMan® Array Human MicroRNA A Cards v2.0
          miRNA Pool : Megaplex™ RT Primers, Human Pool A
          FGMIR : TaqManTM Human MicroRNA Array A
          FGMIR : QS TaqMan OpenArray Rod miRNA Plate
          MIRNA : TaqMan® Array Rodent MicroRNA A Card v2.0


          miRBase Alias:

          Human: hsa-miR-183 (v17)
          Mouse: mmu-miR-183 (v17)
          Rat: rno-miR-183 (v18)

          Back To Top

          Related Products

          • TaqMan® Universal Master Mix II, with UNG
          • TaqMan® Universal Master Mix II, no UNG
          • TaqMan® MicroRNA Reverse Transcription Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline