Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12830
          Assay Name: hsa-miR-183-5p
          miRBase Accession Number: MI0000273
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 129774905 - 129775014 [-] on Build GRCh38
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Species: Human, Mouse, Rat, Alligator mississippiensis, Branchiostoma floridae, Callithrix jacchus, Dasypus novemcinctus, Dog, Gadus morhua, Goat, Guinea pig, Horse, Ornithorhynchus anatinus, Pongo pygmaeus, Pteropus alecto, Rabbit, Salmo salar, Taeniopygia guttata, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC12830

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000066, mir-183

          Mature miRNA Sequence:

          UAUGGCACUGGUAGAAUUCACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-183-5p MIMAT0000261
          Mouse mmu-miR-183-5p MIMAT0000212
          Rat rno-miR-183-5p MIMAT0000860
          Alligator mississippiensis ami-miR-183-5p MIMAT0038196
          Branchiostoma floridae bfl-miR-183 MIMAT0009487
          Callithrix jacchus cja-miR-183 MIMAT0039448
          View More Rows
          Dasypus novemcinctus dno-miR-183-5p MIMAT0047701
          Dog cfa-miR-183 MIMAT0006621
          Gadus morhua gmo-miR-183-5p MIMAT0044261
          Goat chi-miR-183 MIMAT0036007
          Guinea pig cpo-miR-183-5p MIMAT0047050
          Horse eca-miR-183 MIMAT0012938
          Ornithorhynchus anatinus oan-miR-183 MIMAT0006805
          Pongo pygmaeus ppy-miR-183 MIMAT0015780
          Pteropus alecto pal-miR-183-5p MIMAT0040144
          Rabbit ocu-miR-183-5p MIMAT0048287
          Salmo salar ssa-miR-183-5p MIMAT0032429
          Taeniopygia guttata tgu-miR-183 MIMAT0025393
          Tupaia chinensis tch-miR-183-5p MIMAT0036573

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-183
          Stem-loop Accession # MI0000273
          Stem-loop Sequence
          CCGCAGAGUGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA
          Chromosome Location Chr. 7 - 129774905 - 129775014 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000261
          miRBase ID: hsa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 7 - 129774905 - 129775014 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-183
          Stem-loop Accession # MI0000225
          Stem-loop Sequence
          CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAG
          Chromosome Location Chr. 6 - 30169668 - 30169737 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000212
          miRBase ID: mmu-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 6 - 30169668 - 30169737 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-183
          Stem-loop Accession # MI0000928
          Stem-loop Sequence
          CCAGAGAGUGUGACUCCUGUCCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCGCGA
          Chromosome Location Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000860
          miRBase ID: rno-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: Chr. 4 - 57463569 - 57463678 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-183
          Stem-loop Accession # MI0029768
          Stem-loop Sequence
          ACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACGCUAUCAGUGAAUUACCAUAGGGCCAUAAACAGAGCAGAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038196
          miRBase ID: ami-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-183
          Stem-loop Accession # MI0010031
          Stem-loop Sequence
          CCACGUGCCCGUUCACGGUCUAUGGCACUGGUAGAAUUCACUGAUCGUAGACACCAGUGAUUCUUCAGUCAGCCAUACACCGUGGCGGGAAACUGAUCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009487
          miRBase ID: bfl-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-183
          Stem-loop Accession # MI0031971
          Stem-loop Sequence
          UGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039448
          miRBase ID: cja-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-183
          Stem-loop Accession # MI0039014
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047701
          miRBase ID: dno-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-183
          Stem-loop Accession # MI0008017
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006621
          miRBase ID: cfa-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-183
          Stem-loop Accession # MI0036163
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAUAGGGCCAUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044261
          miRBase ID: gmo-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-183
          Stem-loop Accession # MI0030654
          Stem-loop Sequence
          CCAGAGAGCGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCGCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036007
          miRBase ID: chi-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-183
          Stem-loop Accession # MI0038671
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047050
          miRBase ID: cpo-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-183
          Stem-loop Accession # MI0012692
          Stem-loop Sequence
          CUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012938
          miRBase ID: eca-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-183
          Stem-loop Accession # MI0006667
          Stem-loop Sequence
          GGAGGGGGAGACCCUCACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAAAACACACUAUCAGUGAAUUACCAAAGGGCCAUAAACAGAGCAGAGAAAGAACCACAAGACUCUCCCAUCUCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006805
          miRBase ID: oan-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-183
          Stem-loop Accession # MI0014848
          Stem-loop Sequence
          CAGAGCGUGACUCCUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCAGUCAGUGAAUUACCGAAGGGCCAUAAACAGAGCAGAGACAGAUCCACGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015780
          miRBase ID: ppy-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-183
          Stem-loop Accession # MI0032516
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040144
          miRBase ID: pal-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-183
          Stem-loop Accession # MI0039322
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048287
          miRBase ID: ocu-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-183-1
          Stem-loop Accession # MI0026544
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGACGGCACACUAUCAGUGAAUUACCAUAGGGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032429
          miRBase ID: ssa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-183-2
          Stem-loop Accession # MI0026545
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAAGGCACACUAUCAGUGAAUUACCAUUGGGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032429
          miRBase ID: ssa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-183-3
          Stem-loop Accession # MI0026546
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAAAGCACACUAUCAGUGAAUUACCAUAGGGCCAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032429
          miRBase ID: ssa-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-183
          Stem-loop Accession # MI0022170
          Stem-loop Sequence
          CUGUUCUGUGUAUGGCACUGGUAGAAUUCACUGUGCAACCCCGCGGUCAGUGAAUUACCAUAGGGCCAUAAACAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025393
          miRBase ID: tgu-miR-183
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-183
          Stem-loop Accession # MI0031236
          Stem-loop Sequence
          UAUGGCACUGGUAGAAUUCACUGUGAACAGUCUCGGUCAGUGAAUUACCGAAGGGCCAUAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036573
          miRBase ID: tch-miR-183-5p
          Mature miRNA Sequence: UAUGGCACUGGUAGAAUUCACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464630_pri
          TaqMan™ Pri-miRNA Assay : Hs03302720_pri
          TaqMan™ Pri-miRNA Assay : Mm03306888_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12830
          Ambion® Pre-miR™ miRNA Precursor : PM12830
          TaqMan™ MicroRNA Assay : 002269
          mirVana® miRNA inhibitor : MH12830
          TaqMan™ Advanced miRNA Assay : 477937_mir
          TaqMan™ Advanced miRNA Assay : rno481330_mir
          TaqMan™ Advanced miRNA Assay : mmu482690_mir


          miRBase Alias:

          Human: hsa-miR-183 (v17)
          Mouse: mmu-miR-183 (v17)
          Rat: rno-miR-183 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline