Search
Search

| Assay Name: | hsa-miR-18b* |
|---|---|
| miRBase Accession Number: | MI0001518 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGCCCUAAAUGCCCCUUCUGGC |
| Species: | Human, Guinea pig, Rabbit |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UGCCCUAAAUGCCCCUUCUGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-18b-3p | MIMAT0004751 |
| Guinea pig | cpo-miR-18b-3p | MIMAT0046862 |
| Rabbit | ocu-miR-18b-3p | MIMAT0048114 |